Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00512801_m1
          See other GSTT4 GE Assays ›
          Gene Symbol:
          Gstt4
          Gene Name:
          glutathione S-transferase, theta 4
          Gene Aliases:
          4930583C14Rik, AI507011
          Chromosome Location:
          Chr.10: 75814943 - 75822563 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00512801_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gstt4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029472.3 NP_083748.3 4 - 5 649 83
          GenBank mRNA AK016340.1 - 4 - 5 633 83
          BC061011.1 - 4 - 5 649 6774382 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002384.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gstt4

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          glutathione transferase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 NM_029472.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 AK016340.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 BC052144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 BC061011.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 NM_029472.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 AK016340.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 BC052144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P432229 BC061011.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108823 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153946 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 NM_029472.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 AK016340.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 BC052144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 BC061011.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 NM_029472.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 AK016340.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 BC052144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P432229 BC061011.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258801 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153946 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07111349 - -
          Whole Mouse Genome G4122A A_52_P519569 NM_029472.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P519569 AK016340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P519569 BC052144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P519569 BC061011.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 NM_029472.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 AK016340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 BC052144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 BC061011.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 NM_029472.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 AK016340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 BC052144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P432229 BC061011.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110112 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08585805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08820299 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08820300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08820305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08776280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08659241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08444430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08806069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08337800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08806073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08271127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08424232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08820305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08585805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08659245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08776280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08871311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08424235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08904256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08806075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258764 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370011 - -
          Affymetrix HT MG-430 PM Array Plate 1428815_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370007 - -
          Mouse Genome 430 2.0 Array 1428815_at - -
          Mouse Genome 430 2.0 Array 1428815_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75886_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75886_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000017068.mm.1 TC1000002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017068.mm.1 NM_029472.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017068.mm.1 NM_029472.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017069.mm.1 TC1000002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017069.mm.1 NM_029472.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017069.mm.1 NM_029472.3 Refseq
          Mouse Expression Set 430 1428815_at - -
          Mouse Expression Set 430 1428815_PM_at - -
          Murine Genome U74v2 Set 165689_PM_at - -
          Murine Genome U74v2 Set 165689_at - -
          Affymetrix HT Mouse Genome 430B Array 1428815_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428815_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 747099 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 540497 - -
          Illumina mouse-6 v1.0 expression beadchip 540497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217133 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217133 - -
          mouse-6 v1.1 (Illumina) scl37810.5.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 540497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37810.5.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217133 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217133 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0