Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00516081_g1
          See other KIFC3 GE Assays ›
          Gene Symbol:
          Kifc3
          Gene Name:
          kinesin family member C3
          Gene Aliases:
          AI325457, BB123200
          Chromosome Location:
          Chr.8: 95099825 - 95202857 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00516081_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kifc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145831.1 NP_001139303.1 7 - 8 839 84
          NM_001145832.1 NP_001139304.1 3 - 4 323 84
          NM_010631.2 NP_034761.3 5 - 6 666 84
          GenBank mRNA AF013118.2 - 1 - 2 302 84
          AK132895.1 - 3 - 4 323 84
          BC070429.1 - 5 - 6 666 6810893 84
          BI737872.1 - 3 - 4 258 84
          D49546.1 - 5 - 6 681 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002739.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kifc3

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          microtubule-based movement
          Golgi organization
          zonula adherens maintenance
          epithelial cell-cell adhesion
          nucleotide binding
          microtubule motor activity
          ATP binding
          microtubule binding
          minus-end-directed microtubule motor activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800021020.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021020.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021020.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021021.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021021.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021021.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021022.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021022.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021022.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021022.mm.1 NM_010631.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021022.mm.1 NM_010631.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021024.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021024.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021024.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021018.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021018.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021018.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021018.mm.1 NM_001145832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021018.mm.1 NM_001145832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021018.mm.1 NM_010631.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021018.mm.1 NM_010631.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021016.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021016.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021016.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021016.mm.1 NM_001145832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021016.mm.1 NM_001145832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021016.mm.1 NM_010631.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021016.mm.1 NM_010631.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021019.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021019.mm.1 NM_001145832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021019.mm.1 NM_001145832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021015.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021015.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021015.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021015.mm.1 NM_001145832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021015.mm.1 NM_001145832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021015.mm.1 NM_010631.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021015.mm.1 NM_010631.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021026.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021026.mm.1 NM_001145831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021026.mm.1 NM_001145831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021026.mm.1 NM_010631.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021026.mm.1 NM_010631.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021025.mm.1 TC0800002739.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800021025.mm.1 NM_010631.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800021025.mm.1 NM_010631.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 762443 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0