Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517422_m1
          See other ASCL3 GE Assays ›
          Gene Symbol:
          Ascl3
          Gene Name:
          achaete-scute complex homolog 3 (Drosophila)
          Gene Aliases:
          Sgn1, bHLHa42
          Chromosome Location:
          Chr.7: 109727465 - 109731732 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517422_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ascl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020051.1 NP_064435.1 1 - 2 52 65
          GenBank mRNA BC104346.1 - 1 - 2 47 40049496 65
          BC104347.1 - 1 - 2 46 40049502 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004079.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC104346.1, BC104347.1, NM_020051.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ascl3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          DNA-binding transcription repressor activity, RNA polymerase II-specific
          DNA binding
          protein binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185553 NM_020051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185553 AB046448.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185553 AJ277605.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892623 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137531 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185553 NM_020051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185553 AB046448.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185553 AJ277605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P237317 NM_020051.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25409722 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137531 - -
          Whole Mouse Genome G4122A A_52_P460800 NM_020051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460800 AB046448.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460800 AB221543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460800 AJ277605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460800 BC104346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237317 NM_020051.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110118 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06630100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06630095 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06630093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25409710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06539591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06365122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06420310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06365124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06559805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25409729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05954549 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566801 - -
          Affymetrix HT MG-430 PM Array Plate 1420780_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10566800 - -
          Mouse Genome 430 2.0 Array 1420780_at - -
          Mouse Genome 430 2.0 Array 1420780_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56787_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56787_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700033696.mm.1 TC0700004079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033696.mm.1 NM_020051.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033696.mm.1 NM_020051.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033695.mm.1 TC0700004079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033695.mm.1 NM_020051.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033695.mm.1 NM_020051.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033697.mm.1 TC0700004079.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033697.mm.1 NM_020051.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033697.mm.1 NM_020051.1 Refseq
          Mouse Expression Set 430 1420780_PM_at - -
          Mouse Expression Set 430 1420780_at - -
          Mouse Genome 430A 2.0 Array 1420780_PM_at - -
          Mouse Genome 430A 2.0 Array 1420780_at - -
          Affymetrix HT Mouse Genome 430A Array 1420780_at - -
          Affymetrix HT Mouse Genome 430A Array 1420780_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 371128 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4570484 - -
          Illumina mouse-6 v1.0 expression beadchip 4570484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2691996 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2691996 - -
          mouse-6 v1.1 (Illumina) scl30831.2.1_250-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30831.2.1_250-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570484 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2691996 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2691996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0