Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00517965_m1
          See other COMMD5 GE Assays ›
          Gene Symbol:
          Commd5
          Gene Name:
          COMM domain containing 5
          Gene Aliases:
          2310065H03Rik, AI854466, D15Ertd81e, Hcarg
          Chromosome Location:
          Chr.15: 76899941 - 76901297 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00517965_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Commd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025536.2 NP_079812.1 1 - 2 49 59
          GenBank mRNA AK010045.1 - 1 - 2 50 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000613.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK010045.1, NM_025536.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Commd5

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073607 NM_025536.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073607 AK010045.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2073607 AK013148.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 752735 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141546 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073607 NM_025536.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073607 AK010045.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2073607 AK013148.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430485 NM_025536.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430485 AK010045.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430485 AK013148.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430485 BC025891.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351071 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141546 - -
          Whole Mouse Genome G4122A A_51_P430485 NM_025536.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430485 AK010045.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430485 AK013148.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430485 BC025891.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126850 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141546 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12371346 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12371345 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12008379 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12017049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12407844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12008379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12017049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12371346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11944426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12407845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425017 - -
          Affymetrix HT MG-430 PM Array Plate 1451369_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419989_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449694_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425016 - -
          Mouse Genome 430 2.0 Array 1451369_PM_at - -
          Mouse Genome 430 2.0 Array 1449694_s_at - -
          Mouse Genome 430 2.0 Array 1419989_PM_at - -
          Mouse Genome 430 2.0 Array 1449694_PM_s_at - -
          Mouse Genome 430 2.0 Array 1419989_at - -
          Mouse Genome 430 2.0 Array 1451369_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66398_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66398_at - -
          Affymetrix Mouse Clariom S Array PSR1500004504.mm.1 TC1500000613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004504.mm.1 NM_025536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004504.mm.1 NM_025536.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004500.mm.1 TC1500000613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004500.mm.1 NM_025536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004500.mm.1 NM_025536.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004503.mm.1 TC1500000613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004503.mm.1 NM_025536.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004503.mm.1 NM_025536.2 Refseq
          Mouse Expression Set 430 1451369_at - -
          Mouse Expression Set 430 1451369_PM_at - -
          Murine Genome U74v2 Set 94945_PM_at - -
          Murine Genome U74v2 Set 94945_at - -
          Mouse Genome 430A 2.0 Array 1451369_PM_at - -
          Mouse Genome 430A 2.0 Array 1451369_at - -
          Affymetrix HT Mouse Genome 430A Array 1449694_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419989_at - -
          Affymetrix HT Mouse Genome 430A Array 1449694_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451369_at - -
          Affymetrix HT Mouse Genome 430A Array 1451369_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419989_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103830035 - -
          Illumina mouse-6 v1.0 expression beadchip 3450100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214793 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214793 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2472066 - -
          mouse-6 v1.1 (Illumina) scl0066398.2_63-S - -
          mouse-6 v1.1 (Illumina) scl47782.2_279-S - -
          Illumina Sentrix mouse-6.v1 bead array 103830035 - -
          Illumina Sentrix mouse-6.v1 bead array 3450100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066398.2_63-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450100 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103830035 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214793 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214793 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0066398.2_63-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0