Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00518789_g1
          See other APOBEC3 GE Assays ›
          Gene Symbol:
          Apobec3
          Gene Name:
          apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3
          Gene Aliases:
          Arp3, BC003314, Cem15, Rfv3
          Chromosome Location:
          Chr.15: 79892408 - 79908905 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00518789_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apobec3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160415.1 NP_001153887.1 3 - 4 583 79
          NM_030255.3 NP_084531.2 3 - 4 583 79
          GenBank mRNA AB461446.1 - 3 - 4 474 79
          AB461447.1 - 3 - 4 474 79
          AB461448.1 - 3 - 4 474 79
          AB461449.1 - 3 - 4 474 79
          AK044394.1 - 3 - 4 543 79
          AK049256.1 - 3 - 4 543 79
          AK049998.1 - 537 79
          AK153719.1 - 3 - 4 533 79
          AK154035.1 - 588 79
          AK162766.1 - 1 - 2 220 79
          AK167986.1 - 3 - 4 589 79
          BC003314.1 - 3 - 4 565 3155422 79
          EU707568.1 - 3 - 4 474 79
          EU707570.1 - 3 - 4 474 79
          EU707571.1 - 2 - 3 306 79
          GQ871501.1 - 3 - 4 471 79
          GQ871502.1 - 2 - 3 414 79
          GQ871503.1 - 3 - 4 471 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000688.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apobec3

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          immune system process
          metabolic process
          negative regulation of transposition
          negative regulation of viral genome replication
          innate immune response
          defense response to virus
          RNA binding
          catalytic activity
          GTP cyclohydrolase activity
          protein binding
          tRNA-specific adenosine deaminase activity
          zinc ion binding
          hydrolase activity
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds, in cyclic amidines
          N-cyclopropylmelamine deaminase activity
          N-cyclopropylammeline deaminase activity
          N-cyclopropylammelide alkylamino hydrolase activity
          2,5-diamino-6-ribitylamino-4(3H)-pyrimidinone 5'-phosphate deaminase activity
          tRNA-specific adenosine-37 deaminase activity
          archaeal-specific GTP cyclohydrolase activity
          metal ion binding
          deoxycytidine deaminase activity
          tRNA-specific adenosine-34 deaminase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500005139.mm.1 TC1500000688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005139.mm.1 NM_001160415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005139.mm.1 NM_001160415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005139.mm.1 NM_030255.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005139.mm.1 NM_030255.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005138.mm.1 TC1500000688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005138.mm.1 NM_001160415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005138.mm.1 NM_001160415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005138.mm.1 NM_030255.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005138.mm.1 NM_030255.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005140.mm.1 TC1500000688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005140.mm.1 NM_001160415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005140.mm.1 NM_001160415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005140.mm.1 NM_030255.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005140.mm.1 NM_030255.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0