Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00519984_m1
          See other LINCPINT GE Assays ›
          Gene Symbol:
          Lincpint
          Gene Name:
          long intergenic non-protein coding RNA, Trp53 induced transcript
          Gene Aliases:
          MNCb-1768, Pint, linc-Pint
          Chromosome Location:
          Chr.6: 31093563 - 31218474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00519984_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lincpint
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_110470.1 2 - 3 527 65
          GenBank mRNA KC860258.1 - 2 - 3 399 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002013.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism growth
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984259 AB041803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984253 BC145649.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1984253 AB041803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033557 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984259 AB041803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984253 BC145649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1984253 AB041803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033557 - -
          Whole Mouse Genome G4122A A_52_P26265 AB041803.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05046205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05695453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05695455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05578488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05695457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05578477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05578478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05046207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05398533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05578481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05398546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05578480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05575736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05211864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05433146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05578486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05846933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05398553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24336396 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10543786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543789 - -
          Affymetrix HT MG-430 PM Array Plate 1460417_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10543785 - -
          Mouse Genome 430 2.0 Array 1460417_at - -
          Mouse Genome 430 2.0 Array 1458698_PM_at - -
          Mouse Genome 430 2.0 Array 1460417_PM_at - -
          Mouse Genome 430 2.0 Array 1458698_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232685_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232685_at - -
          Affymetrix Mouse Clariom S Array PSR0600015894.mm.1 TC0600002013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015894.mm.1 NR_110469.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015894.mm.1 NR_110470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015896.mm.1 TC0600002013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015896.mm.1 NR_110469.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015896.mm.1 NR_110470.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015896.mm.1 NR_110471.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015895.mm.1 TC0600002013.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015895.mm.1 NR_110469.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015895.mm.1 NR_110470.1 Refseq
          Mouse Expression Set 430 1460417_at - -
          Mouse Expression Set 430 1460417_PM_at - -
          Murine Genome U74v2 Set 134833_at - -
          Murine Genome U74v2 Set 134833_PM_at - -
          Mouse Genome 430A 2.0 Array 1460417_at - -
          Mouse Genome 430A 2.0 Array 1460417_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460417_at - -
          Affymetrix HT Mouse Genome 430A Array 1460417_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214368 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214368 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0