Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00520057_g1
          See other APOA1BP GE Assays ›
          Gene Symbol:
          Apoa1bp
          Gene Name:
          apolipoprotein A-I binding protein
          Gene Aliases:
          AA087124, AI-BP, AIBP, Apoa1ip, ESTM37
          Chromosome Location:
          Chr.3: 88056523 - 88058541 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00520057_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apoa1bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144897.3 NP_659146.1 5 - 6 646 68
          GenBank mRNA AJ344092.1 - 5 - 6 646 68
          AK131615.1 - 5 - 6 857 68
          AK159846.1 - 5 - 6 648 68
          AY566271.1 - 5 - 6 646 68
          BC058362.1 - 5 - 6 644 6819459 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002297.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apoa1bp

          Gene Ontology Categories:

          Function(s) Process(es)

          NADH metabolic process
          NADP metabolic process
          protein homotetramerization
          nucleotide binding
          isomerase activity
          protein homodimerization activity
          metal ion binding
          NADHX epimerase activity
          NADPHX epimerase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22311543 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 AY566271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 BC058362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 NM_144897.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 AJ344092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 AK159846.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 AY566271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 BC058362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 NM_144897.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 AJ344092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P109939 AK159846.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681858 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149916 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142890 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119011 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 AY566271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 BC058362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 NM_144897.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 AJ344092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 AK159846.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 AY566271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 BC058362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 NM_144897.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 AJ344092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P109939 AK159846.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 AY566271.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 BC058362.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 NM_144897.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 AJ344092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 AK159846.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 AY566271.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 BC058362.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 NM_144897.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 AJ344092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P109939 AK159846.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22311545 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142890 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119011 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149916 - -
          Whole Mouse Genome G4122A A_51_P109939 AY566271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 BC058362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 NM_144897.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 AJ344092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 AK159846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 AY566271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 BC058362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 NM_144897.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 AJ344092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P109939 AK159846.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100402 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117860 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119011 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142890 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149916 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02506696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02702097 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02702100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02702096 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02506699 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02506708 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22311543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22311560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02738114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02702100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02506696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02506685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02506709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03185280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02614207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02878095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02761423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22311558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02702097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499315 - -
          Affymetrix HT MG-430 PM Array Plate 1415727_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499309 - -
          Mouse Genome 430 2.0 Array 1415727_at - -
          Mouse Genome 430 2.0 Array 1415727_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246703_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246703_at - -
          Affymetrix Mouse Clariom S Array PSR0300017372.mm.1 TC0300002297.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017372.mm.1 NM_144897.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017372.mm.1 NM_144897.3 Refseq
          Mouse Expression Set 430 1415727_at - -
          Mouse Expression Set 430 1415727_PM_at - -
          Mouse Expression Set 430 1415727_PM_at - -
          Mouse Expression Set 430 1415727_at - -
          Murine Genome U74v2 Set 94032_at - -
          Murine Genome U74v2 Set 94032_PM_at - -
          Murine Genome U74v2 Set 161359_PM_s_at - -
          Murine Genome U74v2 Set 161359_s_at - -
          Mouse Genome 430A 2.0 Array 1415727_PM_at - -
          Mouse Genome 430A 2.0 Array 1415727_at - -
          Affymetrix HT Mouse Genome 430A Array 1415727_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415727_at - -
          Affymetrix HT Mouse Genome 430B Array 1415727_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1415727_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 510576 - -
          Illumina mouse-6 v1.0 expression beadchip 510576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240276 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240276 - -
          mouse-6 v1.1 (Illumina) scl21986.3.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 510576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21986.3.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2997923 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240276 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2997923 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21986.3.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix