Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00520299_m1
          See other TXNDC5 GE Assays ›
          Gene Symbol:
          Txndc5
          Gene Name:
          thioredoxin domain containing 5
          Gene Aliases:
          AL022641, ERp46, PC-TRP
          Chromosome Location:
          Chr.13: 38500266 - 38528824 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00520299_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Txndc5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289598.1 NP_001276527.1 7 - 8 970 77
          NM_001289599.1 NP_001276528.1 7 - 8 937 77
          NM_145367.4 NP_663342.3 7 - 8 994 77
          GenBank mRNA AK136046.1 - 7 - 8 882 77
          AK142049.1 - 7 - 8 980 77
          AK150781.1 - 7 - 8 978 77
          AK169680.1 - 7 - 8 961 77
          AK224911.1 - 6 - 7 647 77
          AK224913.1 - 7 - 8 981 77
          AK224922.1 - 7 - 8 939 77
          AK224923.1 - 7 - 8 961 77
          AK224924.1 - 7 - 8 969 77
          AK224926.1 - 7 - 8 939 77
          AK224927.1 - 7 - 8 990 77
          AK224930.1 - 7 - 8 942 77
          AK224933.1 - 7 - 8 931 77
          AK224934.1 - 7 - 8 985 77
          AK224936.1 - 7 - 8 984 77
          AK224937.1 - 7 - 8 983 77
          AK224938.1 - 7 - 8 978 77
          AK224939.1 - 7 - 8 971 77
          AK224943.1 - 7 - 8 942 77
          AK224945.1 - 7 - 8 978 77
          AK224946.1 - 7 - 8 981 77
          AK224947.1 - 7 - 8 968 77
          AK224948.1 - 7 - 8 942 77
          AK224949.1 - 7 - 8 931 77
          AK224950.1 - 7 - 8 931 77
          AK224951.1 - 7 - 8 989 77
          AK224955.1 - 2 - 3 242 77
          AK224956.1 - 7 - 8 901 77
          AK224957.1 - 7 - 8 947 77
          AK224958.1 - 7 - 8 947 77
          AK224960.1 - 7 - 8 989 77
          AK224962.1 - 7 - 8 949 77
          AK224965.1 - 7 - 8 966 77
          AK224966.1 - 7 - 8 966 77
          AK224967.1 - 7 - 8 957 77
          AK224968.1 - 5 - 6 524 77
          AK224969.1 - 5 - 6 524 77
          AK224970.1 - 7 - 8 931 77
          AK224971.1 - 7 - 8 991 77
          AK224972.1 - 7 - 8 981 77
          AK224973.1 - 7 - 8 931 77
          AK224974.1 - 7 - 8 934 77
          AK224975.1 - 2 - 3 169 77
          AK224976.1 - 7 - 8 964 77
          AK224977.1 - 7 - 8 978 77
          AK224978.1 - 7 - 8 978 77
          AK224979.1 - 7 - 8 965 77
          AK224981.1 - 7 - 8 935 77
          AK224982.1 - 7 - 8 990 77
          AK224983.1 - 7 - 8 979 77
          AK224984.1 - 7 - 8 944 77
          AK224985.1 - 7 - 8 948 77
          AK224986.1 - 7 - 8 992 77
          AK224988.1 - 7 - 8 968 77
          AK224989.1 - 7 - 8 985 77
          AK224990.1 - 7 - 8 980 77
          AK224991.1 - 7 - 8 975 77
          AK224992.1 - 7 - 8 975 77
          AK224993.1 - 7 - 8 975 77
          AK224997.1 - 7 - 8 942 77
          AK224998.1 - 7 - 8 940 77
          AK224999.1 - 7 - 8 962 77
          AK225004.1 - 7 - 8 983 77
          AK225005.1 - 7 - 8 961 77
          AK225007.1 - 7 - 8 983 77
          AK225010.1 - 7 - 8 983 77
          AK225011.1 - 7 - 8 983 77
          AK225012.1 - 7 - 8 752 77
          AK225013.1 - 7 - 8 752 77
          AK225014.1 - 7 - 8 961 77
          AK225015.1 - 7 - 8 752 77
          AK225016.1 - 7 - 8 891 77
          AK225017.1 - 7 - 8 939 77
          AK225019.1 - 7 - 8 971 77
          AK225020.1 - 7 - 8 922 77
          AK225021.1 - 7 - 8 922 77
          AK225022.1 - 7 - 8 770 77
          AK225023.1 - 7 - 8 996 77
          AY243534.1 - 7 - 8 957 77
          AY548112.1 - 7 - 8 918 77
          BC016252.1 - 7 - 8 961 4911455 77
          BC024505.1 - 7 - 8 906 4983496 77
          BC036155.1 - 6 - 7 580 77
          BC046789.1 - 7 - 8 920 5716897 77

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Txndc5

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          response to endoplasmic reticulum stress
          apoptotic cell clearance
          cell redox homeostasis
          protein disulfide isomerase activity
          isomerase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0