Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00520909_m1
          See other OXNAD1 GE Assays ›
          Gene Symbol:
          Oxnad1
          Gene Name:
          oxidoreductase NAD-binding domain containing 1
          Gene Aliases:
          2410002F01Rik, AW492292, BC019806
          Chromosome Location:
          Chr.14: 32085728 - 32103203 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00520909_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oxnad1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145460.2 NP_663435.2 1 - 2 331 81
          GenBank mRNA AK032160.1 - 1 - 2 248 81
          AK139884.1 - 1 - 2 331 81
          BC019806.1 - 1 - 2 109 4018387 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000337.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK139884.1, BC019806.1, NM_145460.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oxnad1

          Gene Ontology Categories:

          Process(es)

          N-ethylmaleimide reductase activity
          oxidoreductase activity
          reduced coenzyme F420 dehydrogenase activity
          sulfur oxygenase reductase activity
          malolactic enzyme activity
          NADPH:sulfur oxidoreductase activity
          epoxyqueuosine reductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021709 AK139884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021709 AK032160.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021709 NM_145460.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021709 AK031978.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36938 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714990 AK030946.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714990 AK081581.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714990 XM_001474597.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714990 XM_001477759.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331903 AK016497.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331903 XM_133663.5 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 331903 XM_907780.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46065 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151798 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021709 AK139884.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021709 AK032160.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021709 NM_145460.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021709 AK031978.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29515540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08074515 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02826632 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02826642 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151798 - -
          Whole Mouse Genome G4122A A_52_P534620 BC019806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534620 AK075924.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534620 AK032160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534620 NM_145460.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534620 AK031978.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106018 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151798 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02826642 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11806671 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11806687 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11806674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29515645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11125253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11539351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11483667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11539349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11135030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11663719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11359320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11135028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11483661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11239686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11807776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29515582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11125248 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10413829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10413831 - -
          Affymetrix HT MG-430 PM Array Plate 1447816_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1426375_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10413826 - -
          Mouse Genome 430 2.0 Array 1447816_PM_x_at - -
          Mouse Genome 430 2.0 Array 1426375_PM_s_at - -
          Mouse Genome 430 2.0 Array 1426375_s_at - -
          Mouse Genome 430 2.0 Array 1447816_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 218885_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 218885_at - -
          Affymetrix Mouse Clariom S Array PSR1400002908.mm.1 TC1400000337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002908.mm.1 NM_145460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002908.mm.1 NM_145460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002912.mm.1 TC1400000337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002912.mm.1 NM_145460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002912.mm.1 NM_145460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002913.mm.1 TC1400000337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002913.mm.1 NM_145460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002913.mm.1 NM_145460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002903.mm.1 TC1400000337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002903.mm.1 NM_145460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002903.mm.1 NM_145460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002909.mm.1 TC1400000337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002909.mm.1 NM_145460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002909.mm.1 NM_145460.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002910.mm.1 TC1400000337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002910.mm.1 NM_145460.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002910.mm.1 NM_145460.2 Refseq
          Mouse Expression Set 430 1426375_s_at - -
          Mouse Expression Set 430 1426375_PM_s_at - -
          Mouse Expression Set 430 1447816_x_at - -
          Mouse Expression Set 430 1447816_PM_x_at - -
          Murine Genome U74v2 Set 161374_f_at - -
          Murine Genome U74v2 Set 161374_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1426375_s_at - -
          Mouse Genome 430A 2.0 Array 1426375_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1426375_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1426375_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1447816_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447816_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 502998 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2650484 - -
          Illumina mouse-6 v1.0 expression beadchip 2650484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769740 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769740 - -
          mouse-6 v1.1 (Illumina) scl0218885.8_148-S - -
          Illumina Sentrix mouse-6.v1 bead array 2650484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0218885.8_148-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650484 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3009558 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3009558 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769740 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0218885.8_148-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0