Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521044_m1
          See other NMRK1 GE Assays ›
          Gene Symbol:
          Nmrk1
          Gene Name:
          nicotinamide riboside kinase 1
          Gene Aliases:
          AA408380, D630020N23Rik, Nrk1
          Chromosome Location:
          Chr.19: 18632016 - 18652191 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521044_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nmrk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145497.2 NP_663472.1 1 - 2 65 63
          GenBank mRNA AK050579.1 - 1 - 2 96 63
          BC016495.1 - 1 - 2 65 4221437 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000320.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK050579.1, BC016495.1, NM_145497.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nmrk1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          phosphorylation
          pyridine nucleotide biosynthetic process
          nucleotide binding
          molecular_function
          ATP binding
          kinase activity
          transferase activity
          metal ion binding
          ribosylnicotinamide kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31992145 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 AK050579.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 AK169962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 BC016495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 NM_145497.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 AK050579.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 AK169962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 BC016495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P663526 NM_145497.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112180 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106163 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121195 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 AK050579.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 AK169962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 BC016495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 NM_145497.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 AK050579.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 AK169962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 BC016495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P663526 NM_145497.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31992146 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121195 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09121637 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09121605 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106163 - -
          Whole Mouse Genome G4122A A_52_P663526 AK050579.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663526 AK169962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663526 BC016495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663526 NM_145497.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663526 AK050579.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663526 AK169962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663526 BC016495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P663526 NM_145497.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126779 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106163 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14368878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14368871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14278279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31992145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31992194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14168274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14473954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14368872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14202746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14168277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14168281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14153139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14470726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14374381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31992318 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461920 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461916 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461910 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461918 - -
          Affymetrix HT MG-430 PM Array Plate 1425646_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448048_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447503_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461909 - -
          Mouse Genome 430 2.0 Array 1447503_PM_at - -
          Mouse Genome 430 2.0 Array 1447503_at - -
          Mouse Genome 430 2.0 Array 1448048_at - -
          Mouse Genome 430 2.0 Array 1425646_at - -
          Mouse Genome 430 2.0 Array 1425646_PM_at - -
          Mouse Genome 430 2.0 Array 1448048_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225994_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225994_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900003205.mm.1 TC1900000320.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003205.mm.1 NM_145497.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003205.mm.1 NM_145497.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003204.mm.1 TC1900000320.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900003204.mm.1 NM_145497.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900003204.mm.1 NM_145497.2 Refseq
          Mouse Expression Set 430 1425646_PM_at - -
          Mouse Expression Set 430 1425646_at - -
          Mouse Expression Set 430 1448048_PM_at - -
          Mouse Expression Set 430 1448048_at - -
          Mouse Expression Set 430 1447503_at - -
          Mouse Expression Set 430 1447503_PM_at - -
          Murine Genome U74v2 Set 95920_PM_at - -
          Murine Genome U74v2 Set 95920_at - -
          Murine Genome U74v2 Set 166971_PM_at - -
          Murine Genome U74v2 Set 166971_at - -
          Mouse Genome 430A 2.0 Array 1425646_PM_at - -
          Mouse Genome 430A 2.0 Array 1448048_at - -
          Mouse Genome 430A 2.0 Array 1448048_PM_at - -
          Mouse Genome 430A 2.0 Array 1425646_at - -
          Affymetrix HT Mouse Genome 430A Array 1448048_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448048_at - -
          Affymetrix HT Mouse Genome 430A Array 1425646_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425646_at - -
          Affymetrix HT Mouse Genome 430B Array 1447503_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447503_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 403690 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3780082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242106 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242106 - -
          mouse-6 v1.1 (Illumina) scl53305.8.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53305.8.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242106 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242106 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0