Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521423_m1
          See other IL17F GE Assays ›
          Gene Symbol:
          Il17f
          Gene Name:
          interleukin 17F
          Gene Aliases:
          C87042, IL-17F
          Chromosome Location:
          Chr.1: 20777146 - 20785274 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521423_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il17f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145856.2 NP_665855.2 2 - 3 319 85
          GenBank mRNA AB116259.1 - 2 - 3 248 85
          AF458064.1 - 1 - 2 224 85
          AY380822.1 - 2 - 3 319 85
          BC145977.1 - 1 - 2 264 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002032.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il17f

          Gene Ontology Categories:

          Function(s) Process(es)

          proteoglycan metabolic process
          inflammatory response
          negative regulation of angiogenesis
          regulation of transforming growth factor beta receptor signaling pathway
          cytokine biosynthetic process
          lymphotoxin A biosynthetic process
          regulation of interleukin-2 biosynthetic process
          regulation of interleukin-6 biosynthetic process
          regulation of interleukin-8 biosynthetic process
          regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          cartilage development
          positive regulation of cytokine production involved in inflammatory response
          cytokine activity
          cytokine receptor binding
          cytokine binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 NM_145856.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 AB116259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 AF458064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 AY380822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 BC145977.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 NM_145856.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 AB116259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 AF458064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 AY380822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P519301 BC145977.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17887 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106614 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 NM_145856.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 AB116259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 AF458064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 AY380822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 BC145977.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 NM_145856.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 AB116259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 AF458064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 AY380822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P519301 BC145977.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04031894 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106614 - -
          Whole Mouse Genome G4122A A_51_P519301 NM_145856.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 AB116259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 AF458064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 AY380822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 BC145977.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 NM_145856.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 AB116259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 AF458064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 AY380822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P519301 BC145977.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118687 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125619 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20074430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00096882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00793406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01050278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00114780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00277752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00096888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00117906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00551194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00713699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00117909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01121344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00117911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00575033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00575028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00645022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00117912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00713702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00114785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00575032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00096895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01121340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01121345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00218463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00219138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00114784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00713697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00114787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00047536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00096886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01121350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20074389 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353416 - -
          Affymetrix Mouse Gene 1.1 ST Array 10353415 - -
          Affymetrix Mouse Clariom S Array PSR0100016609.mm.1 NM_145856.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016609.mm.1 NM_145856.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016609.mm.1 TC0100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016606.mm.1 NM_145856.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016606.mm.1 NM_145856.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016606.mm.1 TC0100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016608.mm.1 NM_145856.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016608.mm.1 NM_145856.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016608.mm.1 TC0100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016605.mm.1 NM_145856.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016605.mm.1 NM_145856.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016605.mm.1 TC0100002032.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016604.mm.1 NM_145856.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016604.mm.1 NM_145856.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016604.mm.1 TC0100002032.mm.1 MTA TC
          Murine Genome U74v2 Set 133331_PM_at - -
          Murine Genome U74v2 Set 133331_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 822787 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 670577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220484 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220484 - -
          mouse-6 v1.1 (Illumina) scl16953.2.1_157-S - -
          Illumina Sentrix mouse-6.v1 bead array 670577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16953.2.1_157-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2741201 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2741201 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix