Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521545_m1
          See other STAP2 GE Assays ›
          Gene Symbol:
          Stap2
          Gene Name:
          signal transducing adaptor family member 2
          Gene Aliases:
          AW049765, STAP-2
          Chromosome Location:
          Chr.17: 55997074 - 56005643 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521545_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145934.2 NP_666046.1 9 - 10 908 82
          NR_102313.1 8 - 9 1012 82
          GenBank mRNA AK050131.1 - 9 - 10 888 82
          AK050172.1 - 8 - 9 980 82
          BC026642.1 - 9 - 10 882 4986113 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002308.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stap2

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253267 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253228 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 AK050131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 NM_145934.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 AK050172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 BC026642.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 AK050131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 NM_145934.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 AK050172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P388847 BC026642.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637227 AK028989.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637227 AK053840.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637227 AK209756.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637227 BC052736.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637227 BC060669.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637227 NM_009238.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637227 X70298.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129022 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127056 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 AK050131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 NM_145934.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 AK050172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 BC026642.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 AK050131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 NM_145934.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 AK050172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P388847 BC026642.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 AK050131.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 NM_145934.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 AK050172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 BC026642.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 AK050131.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 NM_145934.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 AK050172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P388847 BC026642.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31253250 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03338777 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127056 - -
          Whole Mouse Genome G4122A A_51_P388847 AK050131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388847 NM_145934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388847 AK050172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388847 BC026642.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388847 AK050131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388847 NM_145934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388847 AK050172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P388847 BC026642.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103056 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13300375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13300374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13212389 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13300381 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13300383 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13300376 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13171076 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13212393 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13212390 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253267 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31253226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31253286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13573994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13212390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13501042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13501043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31253229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13105396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13577940 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451905 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451894 - -
          Affymetrix HT MG-430 PM Array Plate 1451261_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1424148_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451893 - -
          Mouse Genome 430 2.0 Array 1424148_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451261_PM_s_at - -
          Mouse Genome 430 2.0 Array 1424148_a_at - -
          Mouse Genome 430 2.0 Array 1451261_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106766_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106766_at - -
          Affymetrix Mouse Clariom S Array PSR1700021530.mm.1 TC1700002308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021530.mm.1 NM_145934.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021530.mm.1 NR_102313.1 Refseq
          Murine Genome U74v2 Set 167310_r_at - -
          Murine Genome U74v2 Set 167310_PM_r_at - -
          Affymetrix HT Mouse Genome 430A Array 1424148_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451261_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451261_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424148_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 448315 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6550348 - -
          Illumina mouse-6 v1.0 expression beadchip 6550348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2758562 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2758562 - -
          mouse-6 v1.1 (Illumina) scl49779.7.1_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49779.7.1_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2834687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2758562 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2834687 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2758562 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0