Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00521962_g1
          See other MASP2 GE Assays ›
          Gene Symbol:
          Masp2
          Gene Name:
          mannan-binding lectin serine peptidase 2
          Gene Aliases:
          MASP-2, MAp19
          Chromosome Location:
          Chr.4: 148594877 - 148615482 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00521962_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Masp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001003893.2 NP_001003893.1 2 - 3 279 56
          NM_010767.3 NP_034897.1 2 - 3 279 56
          GenBank mRNA AA238260.1 - 2 - 3 267 56
          AB009459.1 - 2 - 3 269 56
          AI529228.1 - 2 - 3 280 56
          AJ250369.1 - 2 - 3 268 56
          AK050052.1 - 2 - 3 256 56
          AK149421.1 - 1 - 2 333 56
          BC013893.1 - 1 - 2 253 4211293 56
          BF233497.1 - 2 - 3 277 56
          Y19160.1 - 1 - 2 326 56
          Y19163.1 - 1 - 2 296 56

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK149421.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Masp2

          Gene Ontology Categories:

          Function(s) Process(es)

          complement activation, lectin pathway
          immune system process
          proteolysis
          complement activation
          complement activation, classical pathway
          innate immune response
          complement component C4b binding
          serine-type endopeptidase activity
          calcium ion binding
          protein binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          metal ion binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400015871.mm.1 TC0400001935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015871.mm.1 NM_001003893.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015871.mm.1 NM_001003893.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015871.mm.1 NM_010767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015871.mm.1 NM_010767.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015872.mm.1 TC0400001935.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015872.mm.1 NM_001003893.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015872.mm.1 NM_001003893.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015872.mm.1 NM_010767.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015872.mm.1 NM_010767.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550623 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0