Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523117_g1
          See other VASH2 GE Assays ›
          Gene Symbol:
          Vash2
          Gene Name:
          vasohibin 2
          Gene Aliases:
          B130052G07Rik
          Chromosome Location:
          Chr.1: 190947646 - 190979303 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523117_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vash2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027352.1 1 - 2 166 73
          GenBank mRNA AK045261.1 - 1 - 2 95 73
          AK078596.1 - 1 - 2 167 73
          AK163193.1 - 1 - 2 75 73
          BC024141.1 - 1 - 2 69 5149750 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY150367.1 - 1 - 2 75 Chr. 1 - 190978324 - 190979204 [-] on Build GRCm38 73
          GenBankmRNA BY072562.1 - 1 - 2 75 Chr. 1 - 190978289 - 190979204 [-] on Build GRCm38 73
          GenBankmRNA BB650651.1 - 1 - 2 63 Chr. 1 - 190966247 - 190979192 [-] on Build GRCm38 73
          GenBankmRNA BY789774.1 - 1 - 2 99 Chr. 1 - 190978438 - 190979227 [-] on Build GRCm38 73
          GenBankmRNA BQ771226.1 - 1 - 2 140 Chr. 1 - 190978181 - 190979270 [-] on Build GRCm38 73
          GenBankmRNA BB643496.1 - 92 Chr. - - [-] on Build 73
          GenBankmRNA BQ042184.1 - 1 - 2 60 Chr. 1 - 190960447 - 190979190 [-] on Build GRCm38 73
          GenBankmRNA CF736308.1 - 1 - 2 74 Chr. 1 - 190960401 - 190979204 [-] on Build GRCm38 73
          GenBankmRNA BQ745599.1 - 1 - 2 85 Chr. 1 - 190966198 - 190979215 [-] on Build GRCm38 73
          GenBankmRNA BB623771.1 - 1 - 2 161 Chr. 1 - 190966958 - 190979291 [-] on Build GRCm38 73
          GenBankmRNA BU612377.1 - 1 - 2 59 Chr. 1 - 190960446 - 190979189 [-] on Build GRCm38 73
          GenBankmRNA CN694582.1 - 1 - 2 54 Chr. 1 - 190978181 - 190979184 [-] on Build GRCm38 73
          GenBankmRNA CJ174856.1 - 1 - 2 158 Chr. 1 - 190978301 - 190979288 [-] on Build GRCm38 73
          GenBankmRNA BB618765.1 - 1 - 2 85 Chr. 1 - 190972900 - 190979214 [-] on Build GRCm38 73
          GenBankmRNA BY072490.1 - 1 - 2 75 Chr. 1 - 190978272 - 190979204 [-] on Build GRCm38 73
          GenBankmRNA BB640750.1 - 1 - 2 75 Chr. 1 - 190966914 - 190979204 [-] on Build GRCm38 73
          GenBankmRNA CF736588.1 - 1 - 2 71 Chr. 1 - 190960431 - 190979201 [-] on Build GRCm38 73
          GenBankmRNA BY125060.1 - 1 - 2 85 Chr. 1 - 190978340 - 190979214 [-] on Build GRCm38 73
          GenBankmRNA BY775652.1 - 1 - 2 85 Chr. 1 - 190978308 - 190979214 [-] on Build GRCm38 73
          GenBankmRNA BY141587.1 - 1 - 2 98 Chr. 1 - 190978347 - 190979227 [-] on Build GRCm38 73
          GenBankmRNA BU701528.1 - 1 - 2 67 Chr. 1 - 190960431 - 190979197 [-] on Build GRCm38 73
          GenBankmRNA CN697507.1 - 1 - 2 76 Chr. 1 - 190970453 - 190979206 [-] on Build GRCm38 73
          GenBankmRNA CN667706.1 - 1 - 2 148 Chr. 1 - 190978214 - 190979278 [-] on Build GRCm38 73
          GenBankmRNA BY150057.1 - 1 - 2 92 Chr. 1 - 190978383 - 190979222 [-] on Build GRCm38 73
          GenBankmRNA BY133108.1 - 1 - 2 75 Chr. 1 - 190978265 - 190979204 [-] on Build GRCm38 73
          GenBankmRNA BM963403.1 - 1 - 2 61 Chr. 1 - 190960349 - 190979190 [-] on Build GRCm38 73
          GenBankmRNA BY130053.1 - 1 - 2 85 Chr. 1 - 190978383 - 190979214 [-] on Build GRCm38 73
          GenBankmRNA BY012402.1 - 1 - 2 91 Chr. 1 - 190978288 - 190979221 [-] on Build GRCm38 73
          GenBankmRNA BY082830.1 - 1 - 2 138 Chr. 1 - 190978345 - 190979267 [-] on Build GRCm38 73
          GenBankmRNA BY145418.1 - 1 - 2 70 Chr. 1 - 190978318 - 190979199 [-] on Build GRCm38 73
          GenBankmRNA BY130052.1 - 1 - 2 85 Chr. 1 - 190978384 - 190979214 [-] on Build GRCm38 73
          GenBankmRNA CF736795.1 - 1 - 2 59 Chr. 1 - 190966198 - 190979189 [-] on Build GRCm38 73
          GenBankmRNA BY766477.1 - 1 - 2 75 Chr. 1 - 190978286 - 190979204 [-] on Build GRCm38 73
          GenBankmRNA BU701224.1 - 1 - 2 73 Chr. 1 - 190960431 - 190979203 [-] on Build GRCm38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream angel homolog 2 (Drosophila) Angel2 2610307I21Rik,
          5730410O10Rik,
          AI845692,
          D1Ertd396e,
          D1Ertd654e
          -31.2kb Anti-sense Chr. 1 - 190925108 - 190946962 on NCBI Build GRCm38
          Downstream major facilitator superfamily domain containing 7B Mfsd7b 9630055N22Rik,
          FLVCR,
          Flvcr1
          26.5kb Sense Chr. 1 - 191005832 - 191026215 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003796.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK045261.1, AK078596.1, AK163193.1, BC024141.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of endothelial cell proliferation
          regulation of angiogenesis
          positive regulation of angiogenesis
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100031044.mm.1 TC0100003796.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100031044.mm.1 NR_027352.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031044.mm.1 NR_027352.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031045.mm.1 TC0100003796.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100031045.mm.1 NR_027352.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031045.mm.1 NR_027352.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 510238 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0