Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00523599_g1
          See other 5430435G22RIK GE Assays ›
          Gene Symbol:
          5430435G22Rik
          Gene Name:
          RIKEN cDNA 5430435G22 gene
          Gene Aliases:
          Rab7b
          Chromosome Location:
          Chr.1: 131688680 - 131715441 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00523599_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5430435G22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145509.2 NP_663484.1 3 - 4 332 78
          GenBank mRNA AB232641.1 - 2 - 3 176 78
          AK030688.1 - 3 - 4 332 78
          AK036056.1 - 3 - 4 321 78
          AK079999.1 - 3 - 4 300 78
          CJ137398.1 - 3 - 4 251 78
          JX431303.1 - 2 - 3 176 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001173.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 5430435G22Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          small GTPase mediated signal transduction
          endosome to lysosome transport
          protein transport
          Rab protein signal transduction
          positive regulation of interleukin-6 production
          negative regulation of toll-like receptor 4 signaling pathway
          negative regulation of toll-like receptor 9 signaling pathway
          late endosome to Golgi transport
          positive regulation of megakaryocyte differentiation
          positive regulation of NF-kappaB transcription factor activity
          phagosome-lysosome fusion
          nucleotide binding
          GTPase activity
          GTP binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100009471.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009471.mm.1 NM_145509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009470.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009470.mm.1 NM_145509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009475.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009475.mm.1 NM_145509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009467.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009467.mm.1 NM_145509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009472.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009472.mm.1 NM_145509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009466.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009466.mm.1 NM_145509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009473.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009473.mm.1 NM_145509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009474.mm.1 TC0100001173.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009474.mm.1 NM_145509.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix