Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00524081_g1
          See other ZFP7 GE Assays ›
          Gene Symbol:
          Zfp7
          Gene Name:
          zinc finger protein 7
          Gene Aliases:
          KRAB20, KRAB7, Krox-2, Zfp-7, Zfp65, Zfp80, Zfp86-rs1, mszf73-2
          Chromosome Location:
          Chr.15: 76879217 - 76892395 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00524081_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145916.2 NP_666021.2 4 - 5 366 60
          GenBank mRNA AK144887.1 - 4 - 5 578 60
          AK168948.1 - 4 - 5 321 60
          BC011501.1 - 4 - 5 384 3584546 60
          BU613201.1 - 4 - 5 363 60
          U29499.1 - 2 - 3 163 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000612.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp7

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          DNA-binding transcription factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351003 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 AK168948.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 BC011501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 BC062103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 AK144887.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 NM_145916.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 AK168948.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 BC011501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 BC062103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 AK144887.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P240594 NM_145916.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105457 AK168948.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105457 BC011501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105457 BC062103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105457 AK144887.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105457 NM_145916.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 423022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849176 AK016962.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455649 AK054483.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95531 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106561 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141242 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 AK168948.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 BC011501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 BC062103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 AK144887.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 NM_145916.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 AK168948.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 BC011501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 BC062103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 AK144887.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P240594 NM_145916.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105457 AK168948.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105457 BC011501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105457 BC062103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105457 AK144887.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105457 NM_145916.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30351006 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106561 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141242 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08423625 - -
          Whole Mouse Genome G4122A A_52_P289204 AB010310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P588997 BC062103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P588997 AK143414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662183 U29514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662183 AK168948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662183 BC011501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662183 BC062103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662183 AK144887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 AK168948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 BC011501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 BC062103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 AK144887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 NM_145916.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 AK168948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 BC011501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 BC062103.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 AK144887.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240594 NM_145916.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P415305 U29499.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117694 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135828 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141242 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12186356 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351004 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30351003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30351032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30350963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12372068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12271787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12186366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12186358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12065163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12186368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30350999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30351018 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425014 - -
          Affymetrix HT MG-430 PM Array Plate 1451323_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425012 - -
          Mouse Genome 430 2.0 Array 1451323_PM_at - -
          Mouse Genome 430 2.0 Array 1451323_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223669_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223669_at - -
          Affymetrix Mouse Clariom S Array PSR1500004495.mm.1 TC1500000612.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004495.mm.1 NM_145916.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004495.mm.1 NM_145916.2 Refseq
          Murine Genome U74v2 Set 165741_PM_r_at - -
          Murine Genome U74v2 Set 165741_r_at - -
          Affymetrix HT Mouse Genome 430A Array 1451323_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451323_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555855 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2640746 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002640746 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2463901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2463901 - -
          mouse-6 v1.1 (Illumina) scl47783.4_206-S - -
          Illumina Sentrix mouse-6.v1 bead array 2640746 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47783.4_206-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2640746 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2463901 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2463901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0