Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00545830_g1
          See other ALOX12 GE Assays ›
          Gene Symbol:
          Alox12
          Gene Name:
          arachidonate 12-lipoxygenase
          Gene Aliases:
          9930022G08Rik, Alox12p, P-12LO
          Chromosome Location:
          Chr.11: 70241455 - 70255386 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00545830_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alox12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007440.4 NP_031466.2 4 - 5 614 66
          GenBank mRNA AK036898.1 - 4 - 5 612 66
          AK036960.1 - 4 - 5 570 66
          AK079232.1 - 4 - 5 615 66
          AK087724.1 - 4 - 5 615 66
          AK172216.1 - 4 - 5 612 66
          AK172488.1 - 4 - 5 615 66
          BC152329.1 - 4 - 5 619 40141477 66
          BC152403.1 - 4 - 5 619 40141482 66
          S77511.1 - 2 - 3 171 66
          S80446.1 - 4 - 5 626 66
          U04334.1 - 4 - 5 545 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003090.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Alox12

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          movement of cell or subcellular component
          positive regulation of cell population proliferation
          positive regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of muscle cell apoptotic process
          positive regulation of cell death
          arachidonic acid metabolic process
          lipoxygenase pathway
          fatty acid oxidation
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of endothelial cell differentiation
          positive regulation of angiogenesis
          positive regulation of cell adhesion
          positive regulation of smooth muscle cell proliferation
          hepoxilin biosynthetic process
          positive regulation of mitochondrial depolarization
          establishment of skin barrier
          negative regulation of platelet aggregation
          lipoxin A4 biosynthetic process
          lipoxin B4 biosynthetic process
          arachidonate 12(S)-lipoxygenase activity
          iron ion binding
          juvenile hormone epoxide hydrolase activity
          linoleate 13S-lipoxygenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
          hydrolase activity
          epoxide hydrolase B activity
          phenanthrene-9,10-epoxide hydrolase (9R,10R-forming) activity
          epoxide hydrolase A activity
          phenanthrene-3,4-epoxide hydrolase activity
          phenanthrene-1,2-epoxide hydrolase activity
          phenanthrene-9,10-epoxide hydrolase (9S,10S-forming) activity
          phenanthrene-epoxide hydrolase activity
          phenanthrene-9,10-epoxide hydrolase activity
          pyrene-4,5-epoxide hydrolase activity
          metal ion binding
          dioxygenase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100028523.mm.1 TC1100003090.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028523.mm.1 NM_007440.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0