Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00545877_m1
          See other APC GE Assays ›
          Gene Symbol:
          Apc
          Gene Name:
          adenomatosis polyposis coli
          Gene Aliases:
          AI047805, AU020952, AW124434, CC1, Min, mAPC
          Chromosome Location:
          Chr.18: 34220307 - 34322190 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00545877_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007462.3 NP_031488.2 10 - 11 1619 61
          GenBank mRNA AK045053.1 - 10 - 11 1590 61
          AK046280.1 - 10 - 11 1609 61
          AK080907.1 - 10 - 11 1619 61
          BC141141.1 - 9 - 10 1332 9055753 61
          M88127.1 - 9 - 10 1308 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000282.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Mouse Tumor Metastasis Panel Mouse Wnt Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apc

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          kidney development
          hair follicle development
          protein complex assembly
          cellular response to DNA damage stimulus
          negative regulation of microtubule depolymerization
          cell cycle arrest
          metaphase/anaphase transition of mitotic cell cycle
          mitotic spindle assembly checkpoint
          establishment or maintenance of cell polarity
          pattern specification process
          axonogenesis
          negative regulation of cell proliferation
          axis specification
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          Wnt signaling pathway
          cell migration
          stem cell population maintenance
          negative regulation of Wnt signaling pathway
          regulation of cell migration
          positive regulation of cell migration
          regulation of epithelial cell differentiation
          positive regulation of epithelial cell differentiation
          positive regulation of microtubule polymerization
          cytoplasmic microtubule organization
          neuron projection development
          positive regulation of pseudopodium assembly
          regulation of microtubule-based process
          T cell differentiation in thymus
          somatic stem cell population maintenance
          negative regulation of odontogenesis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of MAPK cascade
          skin development
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          canonical Wnt signaling pathway involved in positive regulation of apoptotic process
          regulation of cell differentiation
          positive regulation of cell differentiation
          regulation of osteoblast differentiation
          regulation of osteoclast differentiation
          positive regulation of protein catabolic process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell adhesion
          muscle cell cellular homeostasis
          thymus development
          negative regulation of epithelial cell proliferation
          regulation of nitrogen compound metabolic process
          chromosome organization
          regulation of cell cycle
          positive regulation of cell division
          regulation of attachment of spindle microtubules to kinetochore
          retina development in camera-type eye
          canonical Wnt signaling pathway
          negative regulation of epithelial cell proliferation involved in prostate gland development
          negative regulation of canonical Wnt signaling pathway
          regulation of glutamate metabolic process
          protease binding
          protein binding
          beta-catenin binding
          microtubule binding
          protein kinase regulator activity
          protein kinase binding
          protein complex binding
          gamma-catenin binding
          microtubule plus-end binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800002032.mm.1 NM_007462.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002032.mm.1 NM_007462.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002032.mm.1 TC1800000282.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0