Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00546124_m1
          See other CLEC10A GE Assays ›
          Gene Symbol:
          Clec10a
          Gene Name:
          C-type lectin domain family 10, member A
          Gene Aliases:
          CD301a, M-ASGP-BP-1, Mgl, Mgl1
          Chromosome Location:
          Chr.11: 70166623 - 70170836 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00546124_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec10a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204252.1 NP_001191181.1 1 - 2 193 69
          NM_010796.3 NP_034926.1 1 - 2 193 69
          GenBank mRNA AK170779.1 - 1 - 2 150 69
          BC014811.1 - 1 - 2 26 4456238 69
          S36676.1 - 1 - 2 42 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000942.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK170779.1,BC014811.1,NM_001204252.1,NM_010796.3,S36676.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec10a

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          connective tissue replacement involved in inflammatory response wound healing
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045055 S36676.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045055 NM_010796.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045055 AK140762.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045055 AK170779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045055 BC014811.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118194 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112552 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045055 S36676.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045055 NM_010796.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045055 AK140762.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045055 AK170779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045055 BC014811.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27899265 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112552 - -
          Whole Mouse Genome G4122A A_51_P141413 S36676.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141413 NM_010796.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141413 AK140762.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141413 AK170779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141413 BC014811.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119851 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112552 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09626433 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09626428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09626422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27899275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09365598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09142399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09365597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09297428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09365592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09626420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09626427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27899285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09260678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09297431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09142397 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377783 - -
          Affymetrix HT MG-430 PM Array Plate 1419605_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377782 - -
          Mouse Genome 430 2.0 Array 1419605_PM_at - -
          Mouse Genome 430 2.0 Array 1419605_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17312_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17312_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100008240.mm.1 NM_001204252.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008240.mm.1 NM_001204252.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008240.mm.1 NM_010796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008240.mm.1 NM_010796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008240.mm.1 TC1100000942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008245.mm.1 NM_001204252.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008245.mm.1 NM_001204252.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008245.mm.1 NM_010796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008245.mm.1 NM_010796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008245.mm.1 TC1100000942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008236.mm.1 NM_001204252.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008236.mm.1 NM_001204252.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008236.mm.1 NM_010796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008236.mm.1 NM_010796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008236.mm.1 TC1100000942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008243.mm.1 NM_001204252.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008243.mm.1 NM_001204252.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008243.mm.1 NM_010796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008243.mm.1 NM_010796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008243.mm.1 TC1100000942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008239.mm.1 NM_001204252.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008239.mm.1 NM_001204252.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008239.mm.1 NM_010796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008239.mm.1 NM_010796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008239.mm.1 TC1100000942.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008238.mm.1 NM_001204252.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008238.mm.1 NM_001204252.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008238.mm.1 NM_010796.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008238.mm.1 NM_010796.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008238.mm.1 TC1100000942.mm.1 MTA TC
          Mouse Expression Set 430 1419605_at - -
          Mouse Expression Set 430 1419605_PM_at - -
          Mouse Genome 430A 2.0 Array 1419605_at - -
          Mouse Genome 430A 2.0 Array 1419605_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419605_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419605_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 497255 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249486 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249486 - -
          mouse-6 v1.1 (Illumina) scl41342.11.1_81-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41342.11.1_81-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249486 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249486 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0