Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00549443_g1
          See other FOXRED1 GE Assays ›
          Gene Symbol:
          Foxred1
          Gene Name:
          FAD-dependent oxidoreductase domain containing 1
          Gene Aliases:
          BC024806, TEG-23, Tex23
          Chromosome Location:
          Chr.9: 35204208 - 35211055 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00549443_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Foxred1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291448.1 NP_001278377.1 9 - 10 1272 76
          NM_001291449.1 NP_001278378.1 9 - 10 1277 76
          NM_172291.2 NP_758495.1 9 - 10 1272 76
          GenBank mRNA AK144759.1 - 9 - 10 1198 76
          AK163722.1 - 9 - 10 1131 76
          BC017604.1 - 9 - 10 1230 76
          BC024806.1 - 9 - 10 1142 5361053 76
          BC027386.1 - 6 - 7 1751 76
          BC028892.1 - 7 - 8 2243 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002009.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Foxred1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial respiratory chain complex I assembly
          molecular_function
          N-ethylmaleimide reductase activity
          oxidoreductase activity
          reduced coenzyme F420 dehydrogenase activity
          sulfur oxygenase reductase activity
          malolactic enzyme activity
          NADPH:sulfur oxidoreductase activity
          epoxyqueuosine reductase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26421275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26421276 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC027386.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC028892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 AK163722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC017604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC024806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC027386.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC028892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 AK163722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC017604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254583 BC024806.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 188267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582406 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 497223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140331 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156874 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC027386.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC028892.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 AK163722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC017604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC024806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC027386.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC028892.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 AK163722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC017604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254583 BC024806.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26421255 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156874 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06749345 - -
          Whole Mouse Genome G4122A A_51_P254583 BC027386.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 BC028892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 AK163722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 BC017604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 BC024806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 BC027386.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 BC028892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 AK163722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 BC017604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254583 BC024806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P346803 BC027386.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P346803 BC028892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P346803 AK163722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P346803 BC017604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P346803 BC024806.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102272 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156874 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07607198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07607150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07607197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07607184 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26421275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26421276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26421182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07656246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07607166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07607213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07836740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07722329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26421267 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592116 - -
          Affymetrix HT MG-430 PM Array Plate 1451354_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10592114 - -
          Mouse Genome 430 2.0 Array 1451354_PM_at - -
          Mouse Genome 430 2.0 Array 1451354_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235169_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235169_at - -
          Affymetrix Mouse Clariom S Array PSR0900016692.mm.1 TC0900002009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900016692.mm.1 NM_172291.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900016693.mm.1 TC0900002009.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900016693.mm.1 NM_172291.2 Refseq
          Mouse Expression Set 430 1451354_PM_at - -
          Mouse Expression Set 430 1451354_at - -
          Murine Genome U74v2 Set 103879_at - -
          Murine Genome U74v2 Set 103879_PM_at - -
          Mouse Genome 430A 2.0 Array 1451354_PM_at - -
          Mouse Genome 430A 2.0 Array 1451354_at - -
          Affymetrix HT Mouse Genome 430A Array 1451354_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451354_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2650239 - -
          MouseWG-6 v2 Expression BeadChip 3940309 - -
          Illumina mouse-6 v1.0 expression beadchip 3940309 - -
          Illumina mouse-6 v1.0 expression beadchip 6520348 - -
          Illumina mouse-6 v1.0 expression beadchip 2940397 - -
          Illumina mouse-6 v1.0 expression beadchip 2650239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711754 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2756957 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659247 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2734017 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734017 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659247 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2756957 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711754 - -
          mouse-6 v1.1 (Illumina) scl0003454.1_20-S - -
          mouse-6 v1.1 (Illumina) scl0235169.2_28-S - -
          mouse-6 v1.1 (Illumina) scl0003528.1_25-S - -
          mouse-6 v1.1 (Illumina) scl36048.5_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520348 - -
          Illumina Sentrix mouse-6.v1 bead array 2940397 - -
          Illumina Sentrix mouse-6.v1 bead array 3940309 - -
          Illumina Sentrix mouse-6.v1 bead array 2650239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36048.5_11-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003454.1_20-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0235169.2_28-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003528.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520348 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650239 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940397 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2711754 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2832884 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237151 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2756957 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2832886 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2756957 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237151 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2832884 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2711754 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2832886 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003528.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0235169.2_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0