Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00550869_g1
          See other EXOSC8 GE Assays ›
          Gene Symbol:
          Exosc8
          Gene Name:
          exosome component 8
          Gene Aliases:
          2310032N20Rik, CIP3, mKIAA4013
          Chromosome Location:
          Chr.3: 54728679 - 54735364 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00550869_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Exosc8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163570.1 NP_001157042.1 10 - 11 1064 73
          NM_027148.3 NP_081424.3 10 - 11 1052 73
          GenBank mRNA AJ505005.1 - 10 - 11 712 73
          AK009584.1 - 10 - 11 724 73
          AK169719.1 - 8 - 9 655 73
          AK209232.1 - 1 - 2 68 73
          AK220227.1 - 10 - 11 1052 73
          BC004769.1 - 7 - 8 2101 73
          BC059089.1 - 10 - 11 1081 6509820 73
          BC067250.1 - 9 - 10 695 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001987.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Exosc8

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          biological_process
          RNA binding
          mRNA 3'-UTR AU-rich region binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149453 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149457 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149458 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149459 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149455 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149454 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149452 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149460 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 AK220227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 BC004769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 BC059089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 BC067250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 AK009584.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 AK220227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 BC004769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 BC059089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 BC067250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P496260 AK009584.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52856 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169020 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 AK220227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 BC004769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 BC059089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 BC067250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 AK009584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 AK220227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 BC004769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 BC059089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 BC067250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P496260 AK009584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P500806 BC004769.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P500806 AK047257.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22149417 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169020 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00645566 - -
          Whole Mouse Genome G4122A A_52_P496260 AK220227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 BC004769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 BC059089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 BC067250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 AK009584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 AK220227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 BC004769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 BC059089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 BC067250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P496260 AK009584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500806 BC004769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500806 AK047257.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103319 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169020 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02398685 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02399779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149452 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149455 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149460 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149453 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149458 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149454 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149459 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22149457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22149400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02808892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02399774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02390982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02399771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02762112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02390981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22149400 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10498178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498175 - -
          Affymetrix HT MG-430 PM Array Plate 1428291_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420115_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10498168 - -
          Mouse Genome 430 2.0 Array 1420115_at - -
          Mouse Genome 430 2.0 Array 1449738_s_at - -
          Mouse Genome 430 2.0 Array 1428291_PM_at - -
          Mouse Genome 430 2.0 Array 1420115_PM_at - -
          Mouse Genome 430 2.0 Array 1449738_PM_s_at - -
          Mouse Genome 430 2.0 Array 1428291_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69639_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69639_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300015116.mm.1 TC0300001987.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300015116.mm.1 NM_001163570.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015116.mm.1 NM_001163570.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015116.mm.1 NM_027148.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300015116.mm.1 NM_027148.3 Refseq
          Mouse Expression Set 430 1428291_at - -
          Mouse Expression Set 430 1428291_PM_at - -
          Murine Genome U74v2 Set 95081_at - -
          Murine Genome U74v2 Set 95081_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420115_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420115_at - -
          Affymetrix HT Mouse Genome 430B Array 1428291_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428291_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 360427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360427 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754369 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754369 - -
          mouse-6 v1.1 (Illumina) scl22158.8.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 360427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22158.8.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360427 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2754369 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2754369 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22158.8.1_12-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline