Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00551060_m1
          See other DMRTC2 GE Assays ›
          Gene Symbol:
          Dmrtc2
          Gene Name:
          doublesex and mab-3 related transcription factor like family C2
          Gene Aliases:
          4933432E21Rik, Dmrt7
          Chromosome Location:
          Chr.7: 24869876 - 24877655 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00551060_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dmrtc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027732.2 NP_082008.1 1 - 2 119 65
          GenBank mRNA AF542046.1 - 1 - 2 116 65
          AK017020.1 - 1 - 2 117 65
          BC100434.1 - 1 - 2 119 30918218 65
          BC138203.1 - 1 - 2 50 8861222 65
          BC145988.1 - 1 - 2 50 40131291 65

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF542046.1, BC100434.1, BC138203.1, BC145988.1, NM_027732.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dmrtc2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          male meiosis I
          spermatid nucleus elongation
          sex differentiation
          cell differentiation
          positive regulation of histone H3-K9 dimethylation
          positive regulation of histone H3-K9 trimethylation
          cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein homodimerization activity
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25006994 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25006993 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25006995 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 BC145988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 AF542046.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 AK017020.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 BC100434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 BC138203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 BC145988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 AF542046.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 AK017020.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 BC100434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487004 BC138203.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402309 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206554 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112701 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 BC145988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 AF542046.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 AK017020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 BC100434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 BC138203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 BC145988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 AF542046.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 AK017020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 BC100434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487004 BC138203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 BC145988.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 AF542046.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 AK017020.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 BC100434.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 BC138203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 BC145988.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 AF542046.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 AK017020.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 BC100434.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P487004 BC138203.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006994 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01485998 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112701 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06142015 - -
          Whole Mouse Genome G4122A A_51_P487004 BC145988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 AF542046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 AK017020.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 BC100434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 BC138203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 BC145988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 AF542046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 AK017020.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 BC100434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487004 BC138203.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103724 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134820 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06625990 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25006994 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25006993 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25006995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06269228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06659702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06659692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06156448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06625987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06625988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06562981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06096862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06424227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06424225 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551018 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551017 - -
          Affymetrix HT MG-430 PM Array Plate 1453872_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551011 - -
          Mouse Genome 430 2.0 Array 1453872_at - -
          Mouse Genome 430 2.0 Array 1453872_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71241_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71241_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002953.mm.1 TC0700000400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002953.mm.1 NM_027732.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002953.mm.1 NM_027732.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002952.mm.1 TC0700000400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002952.mm.1 NM_027732.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002952.mm.1 NM_027732.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002954.mm.1 TC0700000400.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002954.mm.1 NM_027732.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002954.mm.1 NM_027732.2 Refseq
          Mouse Expression Set 430 1453872_at - -
          Mouse Expression Set 430 1453872_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453872_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453872_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214790 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214790 - -
          mouse-6 v1.1 (Illumina) scl32946.9.1_202-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32946.9.1_202-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214790 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214790 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0