Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553051_m1
          See other SLC18A2 GE Assays ›
          Gene Symbol:
          Slc18a2
          Gene Name:
          solute carrier family 18 (vesicular monoamine), member 2
          Gene Aliases:
          1110037L13Rik, 9330105E13, Vmat2
          Chromosome Location:
          Chr.19: 59260746 - 59296012 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553051_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc18a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172523.3 NP_766111.1 2 - 3 269 65
          GenBank mRNA AJ555564.1 - 1 - 2 133 65
          AK033871.1 - 2 - 3 225 65
          AK034368.1 - 1 - 2 490 65
          AK035644.1 - 2 - 3 252 65
          AK041274.1 - 2 - 3 221 65
          BC078449.1 - 2 - 3 184 5705966 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000850.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc18a2

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          transport
          neurotransmitter transport
          serotonin transport
          aging
          locomotory behavior
          response to toxic substance
          post-embryonic development
          organic cation transport
          aminergic neurotransmitter loading into synaptic vesicle
          monoamine transport
          insulin secretion
          glucose homeostasis
          xenobiotic transport
          negative regulation of neurotransmitter transport
          transmembrane transport
          cellular response to xenobiotic stimulus
          ammonium transmembrane transport
          amine transmembrane transporter activity
          serotonin:sodium symporter activity
          protein binding
          enzyme binding
          heat shock protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900007366.mm.1 TC1900000850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007366.mm.1 NM_172523.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007366.mm.1 NM_172523.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007367.mm.1 TC1900000850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007367.mm.1 NM_172523.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007367.mm.1 NM_172523.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007369.mm.1 TC1900000850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007369.mm.1 NM_172523.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007369.mm.1 NM_172523.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007368.mm.1 TC1900000850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007368.mm.1 NM_172523.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007368.mm.1 NM_172523.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 749950 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0