Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00553144_g1
          See other SEMA6D GE Assays ›
          Gene Symbol:
          Sema6d
          Gene Name:
          sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D
          Gene Aliases:
          1110067B02Rik, AA409156, D330011G23, mKIAA1479
          Chromosome Location:
          Chr.2: 124089969 - 124667789 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00553144_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sema6d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290997.1 NP_001277926.1 7 - 8 922 101
          NM_001291000.1 NP_001277929.1 3 - 4 1040 101
          NM_172537.4 NP_766125.2 3 - 4 1040 101
          NM_199238.3 NP_954708.1 3 - 4 1040 101
          NM_199239.3 NP_954709.1 2 - 3 275 101
          NM_199240.3 NP_954710.1 2 - 3 275 101
          NM_199241.3 NP_954711.1 2 - 3 275 101
          GenBank mRNA AB091532.1 - 3 - 4 1040 101
          AB091533.1 - 2 - 3 221 101
          AB091534.1 - 2 - 3 221 101
          AB091535.1 - 2 - 3 221 101
          AB091536.1 - 2 - 3 221 101
          AK031307.1 - 3 - 4 736 101
          AK052232.1 - 3 - 4 448 101
          AK084922.1 - 3 - 4 737 101
          AK122515.1 - 7 - 8 925 101
          BC060680.1 - 3 - 4 334 6406252 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001866.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sema6d

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          nervous system development
          positive regulation of smooth muscle cell migration
          negative regulation of smooth muscle cell migration
          ventricular system development
          cell differentiation
          protein binding
          semaphorin receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200014547.mm.1 TC0200001866.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014547.mm.1 NM_172537.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014547.mm.1 NM_199238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014547.mm.1 NM_199239.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014547.mm.1 NM_199240.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014547.mm.1 NM_199241.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014545.mm.1 TC0200001866.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014545.mm.1 NM_172537.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014545.mm.1 NM_199238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014545.mm.1 NM_199239.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014545.mm.1 NM_199240.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014545.mm.1 NM_199241.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014546.mm.1 TC0200001866.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200014546.mm.1 NM_172537.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014546.mm.1 NM_199238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014546.mm.1 NM_199239.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014546.mm.1 NM_199240.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200014546.mm.1 NM_199241.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0