Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00558170_m1
          See other PYHIN1 GE Assays ›
          Gene Symbol:
          Pyhin1
          Gene Name:
          pyrin and HIN domain family, member 1
          Gene Aliases:
          4930422C14, AI447904, Ifi209, Ifix
          Chromosome Location:
          Chr.1: 173630859 - 173647928 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00558170_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pyhin1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175026.3 NP_778191.1 4 - 5 953 79
          GenBank mRNA AK040884.1 - 4 - 5 724 79
          AK080371.1 - 4 - 5 943 79
          AK150817.1 - 4 - 5 844 79
          BC096384.1 - 4 - 5 907 30752094 79
          BC107379.1 - 4 - 5 787 40056704 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001632.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pyhin1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          cellular response to interferon-beta
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          double-stranded DNA binding
          RNA binding
          transcription factor binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036357 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974178 NM_175026.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035509 AK150817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035509 BC096384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035509 AK080371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035509 NM_175026.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035509 AK076726.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036357 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974178 NM_175026.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035509 AK150817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035509 BC096384.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035509 AK080371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035509 NM_175026.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035509 AK076726.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20838856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20838828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20838831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20838857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20838858 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359269 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04359235 - -
          Whole Mouse Genome G4122A A_52_P193657 BC107379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193657 AK150817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193657 BC096384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193657 AK080371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P193657 NM_175026.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334785 AK150817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334785 BC096384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334785 AK080371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334785 NM_175026.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P334785 AK076726.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563280 BC094916.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113719 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04359269 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04359235 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00158375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00525044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20838857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20838923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20838937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20838831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00158375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00409208 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351877 - -
          Affymetrix HT MG-430 PM Array Plate 1435330_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435331_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351873 - -
          Mouse Genome 430 2.0 Array 1435331_PM_at - -
          Mouse Genome 430 2.0 Array 1435330_at - -
          Mouse Genome 430 2.0 Array 1435330_PM_at - -
          Mouse Genome 430 2.0 Array 1435331_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236312_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236312_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100013589.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013589.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013589.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013591.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013591.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013591.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013584.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013584.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013584.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013585.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013585.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013585.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013587.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013587.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013587.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013590.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013590.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013590.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013592.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013592.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013592.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013582.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013582.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013582.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013583.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013583.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013583.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013586.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013586.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013586.mm.1 NM_175026.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013593.mm.1 TC0100001632.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013593.mm.1 NM_175026.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013593.mm.1 NM_175026.3 Refseq
          Mouse Expression Set 430 1435331_PM_at - -
          Mouse Expression Set 430 1435331_at - -
          Mouse Expression Set 430 1435330_PM_at - -
          Mouse Expression Set 430 1435330_at - -
          Murine Genome U74v2 Set 103615_at - -
          Murine Genome U74v2 Set 103615_PM_at - -
          Mouse Genome 430A 2.0 Array 1435330_PM_at - -
          Mouse Genome 430A 2.0 Array 1435331_PM_at - -
          Mouse Genome 430A 2.0 Array 1435330_at - -
          Mouse Genome 430A 2.0 Array 1435331_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 765518 - -
          ABI Mouse Genome Survey Microarray 834755 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3083424 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3083424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0