Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00599954_m1
          See other PRLR GE Assays ›
          Gene Symbol:
          Prlr
          Gene Name:
          prolactin receptor
          Gene Aliases:
          AI987712, Pr-1, Pr-3, Prlr-rs1
          Chromosome Location:
          Chr.15: 10177238 - 10349190 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00599954_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prlr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253781.1 NP_001240710.1 6 - 7 839 113
          NM_001253782.2 NP_001240711.1 5 - 6 648 113
          NM_011169.5 NP_035299.4 6 - 7 1282 113
          GenBank mRNA AB643813.1 - 4 - 5 608 113
          AK050317.1 - 6 - 7 874 113
          AK052679.1 - 6 - 7 737 113
          AK054381.1 - 6 - 7 1102 113
          AK054551.1 - 6 - 7 1283 113
          AK143335.1 - 6 - 7 969 113
          BC005555.1 - 6 - 7 865 3586308 113
          BC006652.1 - 6 - 7 756 3500076 113
          D10214.1 - 4 - 5 557 113
          L13593.1 - 4 - 5 592 113
          L14811.1 - 4 - 5 534 113
          M22957.1 - 6 - 7 759 113
          M22958.1 - 4 - 5 594 113
          M22959.1 - 6 - 7 760 113
          X73372.1 - 4 - 5 534 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002327.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prlr

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of transmembrane receptor protein tyrosine kinase activity
          receptor signaling pathway via JAK-STAT
          lactation
          regulation of cell adhesion
          regulation of epithelial cell differentiation
          prolactin signaling pathway
          mammary gland epithelial cell differentiation
          prostate gland growth
          mammary gland alveolus development
          mammary gland epithelium development
          cytokine receptor activity
          prolactin receptor activity
          protein binding
          peptide hormone binding
          protein kinase binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500019333.mm.1 TC1500002327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500019333.mm.1 NM_001253781.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019333.mm.1 NM_001253781.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019333.mm.1 NM_001253782.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019333.mm.1 NM_001253782.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019333.mm.1 NM_011169.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019333.mm.1 NM_011169.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000704.mm.1 TC1500002327.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500000704.mm.1 NM_001253781.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000704.mm.1 NM_001253781.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000704.mm.1 NM_001253782.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000704.mm.1 NM_001253782.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000704.mm.1 NM_011169.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500000704.mm.1 NM_011169.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0