Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00616368_m1
          See other MIA2 GE Assays ›
          Gene Symbol:
          Mia2
          Gene Name:
          melanoma inhibitory activity 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 59095800 - 59109130 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00616368_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mia2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177321.2 NP_796295.1 1 - 2 111 67
          GenBank mRNA AF390177.2 - 1 - 2 111 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000502.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mia2

          Gene Ontology Categories:

          Function(s) Process(es)

          cholesterol homeostasis
          triglyceride homeostasis
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P168319 NM_177321.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P168319 AF390177.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P168319 NM_177321.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P168319 AF390177.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649894 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92700 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127896 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122878 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P168319 NM_177321.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P168319 AF390177.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P168319 NM_177321.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P168319 AF390177.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28470226 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122878 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127896 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07635245 - -
          Whole Mouse Genome G4122A A_52_P168319 NM_177321.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P168319 AF390177.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P168319 NM_177321.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P168319 AF390177.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116081 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115629 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127896 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122878 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07635238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09985894 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10309139 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10309136 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09985895 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10309140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09985883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28470267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09799908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10174149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09766631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10118241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09813095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09985887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10174150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10412213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10166608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09967081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09766628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28470233 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10395928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395929 - -
          Affymetrix HT MG-430 PM Array Plate 1451964_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10395925 - -
          Mouse Genome 430 2.0 Array 1451964_PM_at - -
          Mouse Genome 430 2.0 Array 1451964_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 338320_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 338320_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200003483.mm.1 TC1200000502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003483.mm.1 NM_177321.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003483.mm.1 NM_177321.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003482.mm.1 TC1200000502.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003482.mm.1 NM_177321.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003482.mm.1 NM_177321.2 Refseq
          Mouse Expression Set 430 1451964_PM_at - -
          Mouse Expression Set 430 1451964_at - -
          Mouse Genome 430A 2.0 Array 1451964_PM_at - -
          Mouse Genome 430A 2.0 Array 1451964_at - -
          Affymetrix HT Mouse Genome 430A Array 1451964_at - -
          Affymetrix HT Mouse Genome 430A Array 1451964_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 832268 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3130181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694320 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694320 - -
          mouse-6 v1.1 (Illumina) scl43181.4.1_246-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43181.4.1_246-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2694320 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694320 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0