Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009977.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009977.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009977.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009975.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009975.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009975.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009980.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009980.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009980.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009973.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009973.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009973.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009974.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009974.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009974.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009972.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009972.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009972.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009982.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009982.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009982.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009978.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009978.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009978.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009983.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009983.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009983.mm.1 |
NM_178703.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009979.mm.1 |
TC0600001268.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009979.mm.1 |
NM_178703.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009979.mm.1 |
NM_178703.4 |
Refseq |