Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00619097_m1
          See other D2HGDH GE Assays ›
          Gene Symbol:
          D2hgdh
          Gene Name:
          D-2-hydroxyglutarate dehydrogenase
          Gene Aliases:
          AA408776, AA408778, AI325464
          Chromosome Location:
          Chr.1: 93824808 - 93852358 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00619097_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: D2hgdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178882.3 NP_849213.2 5 - 6 828 81
          GenBank mRNA AK088200.1 - 775 81
          AK088554.1 - 775 81
          AK141691.1 - 5 - 6 1058 81
          AK170226.1 - 829 81
          BC023277.1 - 796 4006813 81
          BC117794.2 - 5 - 6 766 40092173 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000947.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene D2hgdh

          Gene Ontology Categories:

          Function(s) Process(es)

          response to manganese ion
          response to zinc ion
          response to cobalt ion
          response to magnesium ion
          cellular protein metabolic process
          response to calcium ion
          catalytic activity
          UDP-N-acetylmuramate dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on CH-OH group of donors
          limonene-1,2-diol dehydrogenase activity
          flavin adenine dinucleotide binding
          (R)-2-hydroxyglutarate dehydrogenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051455 BC117794.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051455 AK170226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051455 BC023277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051455 AK088554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051455 AK141691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2123933 NM_178882.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 BY078983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 CD544497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 BC117794.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 AK170226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 AK088554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 BY078983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 CD544497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 BC117794.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 AK170226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P294123 AK088554.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158728 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 BY078983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 CD544497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 BC117794.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 AK170226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 AK088554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 BY078983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 CD544497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 BC117794.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 AK170226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P294123 AK088554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2123933 NM_178882.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051455 BC117794.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051455 AK170226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051455 BC023277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051455 AK088554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051455 AK141691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P257613 AK170226.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P257613 AK088554.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P257613 AK088200.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 BY078983.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 CD544497.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 BC117794.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 AK170226.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 AK088554.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 BY078983.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 CD544497.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 BC117794.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 AK170226.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P294123 AK088554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257613 AK170226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257613 AK088554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257613 AK088200.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 BY078983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 CD544497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 BC117794.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 AK170226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 AK088554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 BY078983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 CD544497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 BC117794.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 AK170226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P294123 AK088554.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00487515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00487519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32978280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00932496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32978274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00008710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00932497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00487524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32978278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00008708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00398508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01094305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457686 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10348890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348892 - -
          Affymetrix HT MG-430 PM Array Plate 1437840_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1437243_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10348889 - -
          Mouse Genome 430 2.0 Array 1437840_PM_s_at - -
          Mouse Genome 430 2.0 Array 1437840_s_at - -
          Mouse Genome 430 2.0 Array 1437243_PM_at - -
          Mouse Genome 430 2.0 Array 1437243_at - -
          Affymetrix Mouse Clariom S Array PSR0100008080.mm.1 TC0100000947.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100008080.mm.1 NM_178882.4 Refseq
          Mouse Expression Set 430 1437243_PM_at - -
          Mouse Expression Set 430 1437243_at - -
          Mouse Expression Set 430 1437840_PM_s_at - -
          Mouse Expression Set 430 1437840_s_at - -
          Murine Genome U74v2 Set 160883_PM_at - -
          Murine Genome U74v2 Set 160883_at - -
          Murine Genome U74v2 Set 99966_at - -
          Murine Genome U74v2 Set 99966_PM_at - -
          Mouse Genome 430A 2.0 Array 1437243_PM_at - -
          Mouse Genome 430A 2.0 Array 1437243_at - -
          Affymetrix HT Mouse Genome 430A Array 1437243_at - -
          Affymetrix HT Mouse Genome 430A Array 1437243_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437840_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1437840_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633525 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550575 - -
          Illumina mouse-6 v1.0 expression beadchip 5550575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252511 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252511 - -
          mouse-6 v1.1 (Illumina) scl0098314.1_111-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0098314.1_111-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252511 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252511 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0