Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00622164_m1
          See other 9430076C15RIK GE Assays ›
          Gene Symbol:
          9430076C15Rik Creb5
          Gene Name:
          RIKEN cDNA 9430076C15 gene
          cAMP responsive element binding protein 5
          Gene Aliases:
          -
          Crebpa, D430026C09Rik
          Chromosome Location:
          Chr.6: 53287295 - 53397216 on Build GRCm38
          Chromosome Location:
          Chr.6: 53573374 - 53697850 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00622164_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9430076C15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015553.2 2 - 3 150 85
          GenBank mRNA AK034642.1 - 125 85
          AK035031.1 - 2 - 3 176 85
          AK051275.1 - 2 - 3 150 85
          AK166503.1 - 2 - 3 162 85
          Gene Symbol: Creb5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK170766.1 - 130 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000637.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2319468 AK051275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2319468 NR_015553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076826 AK166503.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140901 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125150 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142881 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076826 AK166503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2319468 AK051275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2319468 NR_015553.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01331059 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125150 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05902105 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140901 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01331131 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01331159 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05902073 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05902218 - -
          Whole Mouse Genome G4122A A_52_P364113 AK051275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P364113 NR_015553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P253195 AK035031.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125150 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05902218 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140901 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05902105 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01331059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05509631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05287916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05287909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05517120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05302008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05216997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05361066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05830425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05509622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05092455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05333025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05392198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05853917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05360832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05718543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05410074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05853920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05073253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05697192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05073247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05049996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05805432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05241604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05742417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05208519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05241603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05526568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05287923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05171893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05486261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05207431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05460512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05287911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05221854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05807244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05371448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05143975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05287913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05221858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05595941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05147880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05709185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05791589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05722493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24456011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24455775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588984 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446380_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538338 - -
          Mouse Genome 430 2.0 Array 1446380_PM_at - -
          Mouse Genome 430 2.0 Array 1446380_at - -
          Affymetrix Mouse Clariom S Array PSR0600004877.mm.1 TC0600000637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004878.mm.1 TC0600000637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004876.mm.1 TC0600000637.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004879.mm.1 TC0600000637.mm.1 MTA TC
          Mouse Expression Set 430 1446380_at - -
          Mouse Expression Set 430 1446380_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446380_at - -
          Affymetrix HT Mouse Genome 430B Array 1446380_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 400517 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0