Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00622232_s1
          See other C130071C03RIK GE Assays ›
          Gene Symbol:
          C130071C03Rik
          Gene Name:
          RIKEN cDNA C130071C03 gene
          Gene Aliases:
          2810449C10Rik
          Chromosome Location:
          Chr.13: 83728106 - 83735669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00622232_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C130071C03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK048540.1 - 2936 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Inventoried

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P209944 AK048540.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P209944 AK029460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P209944 AK048540.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P209944 AK029460.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564825 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163830 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P209944 AK048540.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P209944 AK029460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P209944 AK048540.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P209944 AK029460.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07937958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07937965 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163830 - -
          Whole Mouse Genome G4122A A_52_P215409 AK047851.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215409 AK039679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215409 AK013308.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P215409 NR_015561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209944 AK048540.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209944 AK029460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209944 AK048540.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209944 AK029460.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11028461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11040997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11028472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11042837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11041015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10803497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29187545 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453791_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1459684_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435179_PM_at - -
          Mouse Genome 430 2.0 Array 1453791_PM_at - -
          Mouse Genome 430 2.0 Array 1447298_at - -
          Mouse Genome 430 2.0 Array 1459684_at - -
          Mouse Genome 430 2.0 Array 1453791_at - -
          Mouse Genome 430 2.0 Array 1459684_PM_at - -
          Mouse Genome 430 2.0 Array 1435179_PM_at - -
          Mouse Genome 430 2.0 Array 1447298_PM_at - -
          Mouse Genome 430 2.0 Array 1435179_at - -
          Mouse Expression Set 430 1453791_at - -
          Mouse Expression Set 430 1459684_PM_at - -
          Mouse Expression Set 430 1447298_at - -
          Mouse Expression Set 430 1447298_PM_at - -
          Mouse Expression Set 430 1453791_PM_at - -
          Mouse Expression Set 430 1459684_at - -
          Mouse Expression Set 430 1435179_at - -
          Mouse Expression Set 430 1435179_PM_at - -
          Murine Genome U74v2 Set 171586_PM_at - -
          Murine Genome U74v2 Set 171586_at - -
          Murine Genome U74v2 Set 139205_at - -
          Murine Genome U74v2 Set 139205_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453791_at - -
          Affymetrix HT Mouse Genome 430B Array 1459684_at - -
          Affymetrix HT Mouse Genome 430B Array 1435179_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453791_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435179_at - -
          Affymetrix HT Mouse Genome 430B Array 1459684_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507757 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0