Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00623919_s1
          See other SPATA33 GE Assays ›
          Gene Symbol:
          Spata33
          Gene Name:
          spermatogenesis associated 33
          Gene Aliases:
          4732415M23Rik, AI847264
          Chromosome Location:
          Chr.8: 123212816 - 123222045 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00623919_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK028607.1 - 1 1154 86
          BC059919.1 - 1 1102 6827099 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001513.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26238534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26238532 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580533 AK076656.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580533 NM_177279.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580533 AK076656.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P580533 NM_177279.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127266 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148845 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580533 AK076656.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580533 NM_177279.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580533 AK076656.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P580533 NM_177279.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26238536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148845 - -
          Whole Mouse Genome G4122A A_52_P580533 AK076656.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580533 NM_177279.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580533 AK076656.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P580533 NM_177279.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327768 AK028607.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105066 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06718114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06718120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06718112 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06718111 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06718121 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26238534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26238532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26238587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06718110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07043941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07268211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07404242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06718115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07393606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06718109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06737855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06912800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26238542 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576251 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576255 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576252 - -
          Affymetrix HT MG-430 PM Array Plate 1435365_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576249 - -
          Mouse Genome 430 2.0 Array 1435365_at - -
          Mouse Genome 430 2.0 Array 1435365_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320869_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320869_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800012367.mm.1 TC0800001513.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800012369.mm.1 TC0800001513.mm.1 MTA TC
          Mouse Expression Set 430 1435365_at - -
          Mouse Expression Set 430 1435365_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435365_at - -
          Affymetrix HT Mouse Genome 430B Array 1435365_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 846226 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104200332 - -
          Illumina mouse-6 v1.0 expression beadchip 5290025 - -
          Illumina mouse-6 v1.0 expression beadchip 104200332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689818 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213979 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689818 - -
          mouse-6 v1.1 (Illumina) scl0320869.2_188-S - -
          Illumina Sentrix mouse-6.v1 bead array 5290025 - -
          Illumina Sentrix mouse-6.v1 bead array 104200332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0320869.2_188-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290025 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104200332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2962386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2689818 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2962386 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0