Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
TC0100003140.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046167.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046167.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046168.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046168.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046170.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025508.mm.1 |
NR_046170.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025512.mm.1 |
TC0100003140.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025512.mm.1 |
NR_046167.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025512.mm.1 |
NR_046167.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025512.mm.1 |
NR_046168.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025512.mm.1 |
NR_046168.1 |
Refseq |
Mouse Expression Set 430 |
1443574_at |
- |
- |
Mouse Expression Set 430 |
1443574_PM_at |
- |
- |