Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00627179_m1
          See other MYCN GE Assays ›
          Gene Symbol:
          Mycn
          Gene Name:
          v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian)
          Gene Aliases:
          N-myc, Nmyc, Nmyc-1, Nmyc1, bHLHe37, c-nmyc
          Chromosome Location:
          Chr.12: 12936093 - 12941836 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00627179_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mycn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008709.3 NP_032735.3 1 - 2 207 58
          GenBank mRNA AK144126.1 - 1 - 2 207 58
          AK146696.1 - 1 - 2 208 58
          AK146913.1 - 1 - 2 205 58
          BC005453.1 - 1 - 2 91 3495446 58

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK146696.1, AK146913.1, BC005453.1, NM_008709.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mycn

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          positive regulation of mesenchymal cell proliferation
          regulation of transcription, DNA-templated
          positive regulation of cell proliferation
          positive regulation of cell death
          lung development
          embryonic digit morphogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic skeletal system morphogenesis
          negative regulation of astrocyte differentiation
          branching morphogenesis of an epithelial tube
          negative regulation of reactive oxygen species metabolic process
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200010063.mm.1 NM_008709.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010063.mm.1 NM_008709.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010063.mm.1 TC1200001432.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010060.mm.1 NM_008709.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010060.mm.1 NM_008709.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010060.mm.1 TC1200001432.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200010064.mm.1 NM_008709.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010064.mm.1 NM_008709.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200010064.mm.1 TC1200001432.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline