Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00651908_m1
          See other 1700018B24RIK GE Assays ›
          Gene Symbol:
          1700018B24Rik
          Gene Name:
          enhancer of rudimentary homolog pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 48605731 - 48609100 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00651908_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700018B24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006080.1 - 1 - 2 406 151
          AK006613.1 - 1 - 2 151
          AK018885.1 - 1 - 2 375 151
          BC049578.1 - 1 - 2 389 151
          BC061086.1 - 1 - 2 405 151

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 NR_003617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 AK006613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 AK018885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 BC049578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 BC061086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 NR_003617.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 AK006613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 AK018885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 BC049578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P625315 BC061086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110230 NR_003617.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142124 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110230 NR_003617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 NR_003617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 AK006613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 AK018885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 BC049578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 BC061086.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 NR_003617.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 AK006613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 AK018885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 BC049578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P625315 BC061086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276787 NR_003617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276787 AK006080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276787 AK006613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276787 AK018885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276787 BC049578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 NR_003617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 AK006613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 AK018885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 BC049578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 BC061086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 NR_003617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 AK006613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 AK018885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 BC049578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P625315 BC061086.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02773555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02876849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03135032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03122362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22115518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02648283 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10498004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10498003 - -
          Affymetrix HT MG-430 PM Array Plate 1451061_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10498002 - -
          Mouse Genome 430 2.0 Array 1451061_PM_at - -
          Mouse Genome 430 2.0 Array 1451061_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66332_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66332_at - -
          Mouse Expression Set 430 1451061_at - -
          Mouse Expression Set 430 1451061_PM_at - -
          Murine Genome U74v2 Set 165890_at - -
          Murine Genome U74v2 Set 165890_PM_at - -
          Mouse Genome 430A 2.0 Array 1451061_at - -
          Mouse Genome 430A 2.0 Array 1451061_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451061_at - -
          Affymetrix HT Mouse Genome 430A Array 1451061_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 918436 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3990647 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231777 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231777 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1231777 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231777 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0