Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652419_m1
          See other CD209C GE Assays ›
          Gene Symbol:
          Cd209c
          Gene Name:
          CD209c antigen
          Gene Aliases:
          SIGNR2, mSIGNR2
          Chromosome Location:
          Chr.8: 3940221 - 3955947 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00652419_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd209c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130903.3 NP_570973.1 3 - 4 1231 85
          GenBank mRNA AF373410.1 - 3 - 4 1231 85
          BC120804.1 - 3 - 4 421 40129727 85
          BC132635.1 - 3 - 4 421 40130912 85
          BC145088.1 - 3 - 4 421 9053636 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001625.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd209c

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          mannose binding
          carbohydrate binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040805 NM_130903.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040805 AF373410.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050754 BC145088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050754 NM_130903.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050754 AF373410.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050754 BC120804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050754 BC132635.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 571955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83151 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107103 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050754 BC145088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050754 NM_130903.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050754 AF373410.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050754 BC120804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050754 BC132635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040805 NM_130903.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040805 AF373410.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182538 BC145088.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182538 NM_130903.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182538 AF373410.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182538 BC120804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182538 BC132635.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104342 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576832 - -
          Affymetrix HT MG-430 PM Array Plate 1421562_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576829 - -
          Mouse Genome 430 2.0 Array 1421562_at - -
          Mouse Genome 430 2.0 Array 1421562_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170776_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170776_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800013373.mm.1 TC0800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013373.mm.1 NM_130903.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013373.mm.1 NM_130903.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013371.mm.1 TC0800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013371.mm.1 NM_130903.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013371.mm.1 NM_130903.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013372.mm.1 TC0800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800013372.mm.1 NM_130903.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013372.mm.1 NM_130903.3 Refseq
          Mouse Expression Set 430 1421562_PM_at - -
          Mouse Expression Set 430 1421562_at - -
          Mouse Genome 430A 2.0 Array 1421562_PM_at - -
          Mouse Genome 430A 2.0 Array 1421562_at - -
          Affymetrix HT Mouse Genome 430A Array 1421562_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421562_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 495530 - -
          ABI Mouse Genome Survey Microarray 508300 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1570286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001570286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646926 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646926 - -
          mouse-6 v1.1 (Illumina) scl00170776.1_193-S - -
          Illumina Sentrix mouse-6.v1 bead array 1570286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00170776.1_193-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1570286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3054469 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646926 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3131225 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3131225 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646926 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3054469 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0