Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655340_s1
          See other MRGPRA4 GE Assays ›
          Gene Symbol:
          Mrgpra4
          Gene Name:
          MAS-related GPR, member A4
          Gene Aliases:
          Mgrg10, MrgA4
          Chromosome Location:
          Chr.7: 47980794 - 47982296 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655340_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrgpra4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153524.2 NP_705744.2 1 947 106
          GenBank mRNA AY042194.1 - 1 947 106
          BC115741.1 - 1 746 40045594 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003060.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrgpra4

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G protein-coupled receptor signaling pathway
          neuropeptide signaling pathway
          chemical synaptic transmission
          G protein-coupled receptor activity
          neuropeptide receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122660 NM_153524.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122660 AY042194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P122660 BC115741.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198664 AY042194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P198664 AY042194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198664 AY042194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P198664 AY042194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122660 NM_153524.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122660 AY042194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P122660 BC115741.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P198664 AY042194.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P198664 AY042194.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25106040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25106009 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06189026 - -
          Whole Mouse Genome G4122A A_51_P198664 AY042194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P198664 AY042194.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122660 NM_153524.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122660 AY042194.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122660 BC115741.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25106051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25106049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25106011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25106009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25106011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25105991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25106051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25106004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25106049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25106040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25105994 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563735 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563736 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563738 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563739 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563737 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563716 - -
          Affymetrix HT MG-430 PM Array Plate 1451926_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563734 - -
          Mouse Genome 430 2.0 Array 1451926_PM_at - -
          Mouse Genome 430 2.0 Array 1451926_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235854_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235854_at - -
          Affymetrix Mouse Clariom S Array PSR0700027973.mm.1 TC0700003060.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700027973.mm.1 NM_153524.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027973.mm.1 NM_153524.2 Refseq
          Mouse Expression Set 430 1451926_PM_at - -
          Mouse Expression Set 430 1451926_at - -
          Mouse Genome 430A 2.0 Array 1451926_at - -
          Mouse Genome 430A 2.0 Array 1451926_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451926_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451926_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 667900 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2570446 - -
          Illumina mouse-6 v1.0 expression beadchip 2570446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248378 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248378 - -
          mouse-6 v1.1 (Illumina) scl0235854.1_68-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0235854.1_68-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2781430 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2781430 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0