Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00660670_g1
          See other CTC1 GE Assays ›
          Gene Symbol:
          Ctc1
          Gene Name:
          CTS telomere maintenance complex component 1
          Gene Aliases:
          1500010J02Rik, AAF-132, AAF132
          Chromosome Location:
          Chr.11: 69015904 - 69036473 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00660670_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013256.2 NP_001013274.2 13 - 14 2589 66
          NM_001143790.1 NP_001137262.1 13 - 14 2589 66
          NM_001281464.1 NP_001268393.1 13 - 14 2460 66
          NM_001281465.1 NP_001268394.1 13 - 14 2573 66
          GenBank mRNA AK038651.1 - 13 - 14 2376 66
          AK044055.1 - 13 - 14 2368 66
          AK145919.1 - 13 - 14 2404 66
          AK165906.1 - 13 - 14 2389 66
          AK182922.1 - 1 - 2 91 66
          AK197567.1 - 1 - 2 91 66
          AK213842.1 - 1 - 2 40 66
          BC014687.1 - 4 - 5 760 66
          BC060629.1 - 9 - 10 1639 6847619 66
          BC066078.1 - 13 - 14 2403 66
          BC072625.1 - 13 - 14 2417 66
          BC147495.1 - 13 - 14 2387 9056325 66
          BC147497.1 - 13 - 14 2387 9056354 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000891.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctc1

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          cellular response to DNA damage stimulus
          aging
          regulation of G2/M transition of mitotic cell cycle
          telomere maintenance via telomere lengthening
          multicellular organism growth
          positive regulation of DNA replication
          positive regulation of fibroblast proliferation
          spleen development
          thymus development
          bone marrow development
          chromosome organization
          hematopoietic stem cell proliferation
          replicative senescence
          DNA binding
          single-stranded DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100007457.mm.1 TC1100000891.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007457.mm.1 NM_001013256.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007457.mm.1 NM_001013256.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007457.mm.1 NM_001143790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007457.mm.1 NM_001143790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007457.mm.1 NM_001281464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007457.mm.1 NM_001281465.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0