Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00661127_s1
          See other 1700065D16RIK GE Assays ›
          Gene Symbol:
          1700065D16Rik
          Gene Name:
          RIKEN cDNA 1700065D16 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 95855418 - 95857882 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00661127_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700065D16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK045280.1 - 1 2051 107
          BC053441.1 - 1 2258 6332684 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002871.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744299 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744294 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744296 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P112033 BC053441.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P112033 AK045280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P112033 NM_028533.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P112033 BC053441.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P112033 AK045280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P112033 NM_028533.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282787 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155470 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P112033 BC053441.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P112033 AK045280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P112033 NM_028533.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P112033 BC053441.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P112033 AK045280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P112033 NM_028533.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26744299 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155470 - -
          Whole Mouse Genome G4122A A_52_P230601 BC053441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230601 AK045280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230601 AK006889.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230601 NM_028533.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P112033 BC053441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P112033 AK045280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P112033 NM_028533.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P112033 BC053441.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P112033 AK045280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P112033 NM_028533.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117482 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07761515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07734273 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07761514 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07761516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07734264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744299 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744294 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26744297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26744313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07873037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08196283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08085885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07561569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07734266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26744313 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10595790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595788 - -
          Affymetrix HT MG-430 PM Array Plate 1453667_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10595787 - -
          Mouse Genome 430 2.0 Array 1457335_PM_at - -
          Mouse Genome 430 2.0 Array 1438921_PM_at - -
          Mouse Genome 430 2.0 Array 1453667_PM_at - -
          Mouse Genome 430 2.0 Array 1457335_at - -
          Mouse Genome 430 2.0 Array 1453667_at - -
          Mouse Genome 430 2.0 Array 1438921_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73410_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73410_at - -
          Affymetrix Mouse Clariom S Array PSR0900023665.mm.1 TC0900002871.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023665.mm.1 NM_001271569.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900023667.mm.1 TC0900002871.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023660.mm.1 TC0900002871.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023662.mm.1 TC0900002871.mm.1 MTA TC
          Mouse Expression Set 430 1453667_PM_at - -
          Mouse Expression Set 430 1453667_at - -
          Affymetrix HT Mouse Genome 430B Array 1453667_at - -
          Affymetrix HT Mouse Genome 430B Array 1453667_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 374331 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5420129 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636814 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636814 - -
          mouse-6 v1.1 (Illumina) scl35493.1_62-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420129 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35493.1_62-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636814 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636814 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0