Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00712725_g1
          See other GORASP1 GE Assays ›
          Gene Symbol:
          Gorasp1
          Gene Name:
          golgi reassembly stacking protein 1
          Gene Aliases:
          5430411C10Rik, GOLPH5, GRASP65, P65
          Chromosome Location:
          Chr.9: 119925673 - 119937558 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00712725_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gorasp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028976.2 NP_083252.1 8 - 9 1119 80
          GenBank mRNA AK017293.1 - 1 - 2 147 80
          AK154986.1 - 8 - 9 1096 80
          AK170627.1 - 8 - 9 1083 80
          BC012251.1 - 8 - 9 1119 4166619 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003249.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gorasp1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation
          transport
          Golgi organization
          protein transport
          negative regulation of dendrite morphogenesis
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26875190 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125972 BC012251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125972 AK204247.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125972 AK209806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125972 AK213744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125972 AK154986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 BC012251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 NM_028976.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 AK017293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 BC062967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 AK122511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 BC012251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 NM_028976.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 AK017293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 BC062967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P45841 AK122511.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168978 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157375 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125972 BC012251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125972 AK204247.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125972 AK209806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125972 AK213744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125972 AK154986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 BC012251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 NM_028976.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 AK017293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 BC062967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 AK122511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 BC012251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 NM_028976.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 AK017293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 BC062967.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P45841 AK122511.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26875183 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06948089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06948101 - -
          Whole Mouse Genome G4122A A_52_P45841 BC012251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 NM_028976.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 AK017293.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 BC062967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 AK122511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 BC012251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 NM_028976.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 AK017293.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 BC062967.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P45841 AK122511.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140286 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115381 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07942249 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07942243 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07942244 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07782561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07942237 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26875190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26875191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07636486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07636482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07850558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07942244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07665345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07661177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07782561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26875181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10597749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597750 - -
          Affymetrix HT MG-430 PM Array Plate 1451671_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425334_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10597748 - -
          Affymetrix Mouse Gene 1.1 ST Array 10590191 - -
          Mouse Genome 430 2.0 Array 1451671_PM_at - -
          Mouse Genome 430 2.0 Array 1425334_at - -
          Mouse Genome 430 2.0 Array 1451671_at - -
          Mouse Genome 430 2.0 Array 1425334_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74498_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74498_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900027409.mm.1 TC0900003249.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027409.mm.1 NM_028976.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027409.mm.1 NM_028976.2 Refseq
          Mouse Expression Set 430 1425334_PM_at - -
          Mouse Expression Set 430 1451671_at - -
          Mouse Expression Set 430 1451671_PM_at - -
          Mouse Expression Set 430 1425334_at - -
          Mouse Genome 430A 2.0 Array 1451671_PM_at - -
          Mouse Genome 430A 2.0 Array 1425334_PM_at - -
          Mouse Genome 430A 2.0 Array 1425334_at - -
          Mouse Genome 430A 2.0 Array 1451671_at - -
          Affymetrix HT Mouse Genome 430A Array 1425334_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451671_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425334_at - -
          Affymetrix HT Mouse Genome 430A Array 1451671_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 784403 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102190673 - -
          Illumina mouse-6 v1.0 expression beadchip 5570309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570309 - -
          mouse-6 v1.1 (Illumina) scl0074498.2_136-S - -
          mouse-6 v1.1 (Illumina) scl35216.9_534-S - -
          Illumina Sentrix mouse-6.v1 bead array 102190673 - -
          Illumina Sentrix mouse-6.v1 bead array 5570309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0074498.2_136-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102190673 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2962511 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2962511 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235664 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242510 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0