Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600002827.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002827.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002827.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002824.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002824.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002824.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002819.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002819.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002819.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002820.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002820.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002820.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002825.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002825.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002825.mm.1 |
NM_177185.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002822.mm.1 |
TC0600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600002822.mm.1 |
NM_177185.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600002822.mm.1 |
NM_177185.4 |
Refseq |