Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00725486_s1
          See other ATPIF1 GE Assays ›
          Gene Symbol:
          Atpif1
          Gene Name:
          ATPase inhibitory factor 1
          Gene Aliases:
          Atpi, IF(1), If1
          Chromosome Location:
          Chr.4: 132530555 - 132535409 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00725486_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atpif1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF002718.1 - 3 378 116
          AK008346.1 - 3 376 116
          AK152389.1 - 4 1393 116
          AV462016.1 - 3 439 116
          BC012680.1 - 3 437 4217188 116
          CB848021.2 - 3 401 116

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312759 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312750 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312749 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 CB848021.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 AF002718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 AK008346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 AK152389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 BC012680.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 CB848021.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 AF002718.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 AK008346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 AK152389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518488 BC012680.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168815 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104622 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 CB848021.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 AF002718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 AK008346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 AK152389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 BC012680.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 CB848021.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 AF002718.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 AK008346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 AK152389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518488 BC012680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 CB848021.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 AF002718.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 AK008346.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 AK152389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 BC012680.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 CB848021.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 AF002718.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 AK008346.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 AK152389.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518488 BC012680.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23312756 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104622 - -
          Whole Mouse Genome G4122A A_51_P518488 CB848021.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 AF002718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 AK008346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 AK152389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 BC012680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 CB848021.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 AF002718.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 AK008346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 AK152389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518488 BC012680.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P276412 AK011886.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P276412 AK161394.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128740 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103513 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03502775 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03502784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03730591 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03502783 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03502782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312750 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312749 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23312759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23312777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03232950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03502783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03507086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03439830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03502771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04060172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03541044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23312741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03748112 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516945 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516944 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516947 - -
          Affymetrix HT MG-430 PM Array Plate 1453681_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434198_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448770_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516943 - -
          Mouse Genome 430 2.0 Array 1448770_a_at - -
          Mouse Genome 430 2.0 Array 1448770_PM_a_at - -
          Mouse Genome 430 2.0 Array 1453681_at - -
          Mouse Genome 430 2.0 Array 1434198_PM_at - -
          Mouse Genome 430 2.0 Array 1453681_PM_at - -
          Mouse Genome 430 2.0 Array 1434198_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11983_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11983_PM_at - -
          Mouse Expression Set 430 1448770_a_at - -
          Mouse Expression Set 430 1434198_PM_at - -
          Mouse Expression Set 430 1434198_at - -
          Mouse Expression Set 430 1448770_PM_a_at - -
          Mouse Expression Set 430 1453681_PM_at - -
          Mouse Expression Set 430 1453681_at - -
          Murine Genome U74v2 Set 93984_PM_at - -
          Murine Genome U74v2 Set 93984_at - -
          Mouse Genome 430A 2.0 Array 1448770_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1448770_a_at - -
          Mouse Genome 430A 2.0 Array 1434198_PM_at - -
          Mouse Genome 430A 2.0 Array 1434198_at - -
          Affymetrix HT Mouse Genome 430A Array 1434198_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434198_at - -
          Affymetrix HT Mouse Genome 430A Array 1448770_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448770_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1453681_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453681_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 462444 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102370091 - -
          Illumina mouse-6 v1.0 expression beadchip 102370091 - -
          Illumina mouse-6 v1.0 expression beadchip 7050577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250068 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252120 - -
          mouse-6 v1.1 (Illumina) scl0011983.1_209-S - -
          Illumina Sentrix mouse-6.v1 bead array 102370091 - -
          Illumina Sentrix mouse-6.v1 bead array 7050577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0011983.1_209-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102370091 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1250068 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250068 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252120 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0011983.1_209-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0