Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00770070_g1
          See other RRP9 GE Assays ›
          Gene Symbol:
          Rrp9
          Gene Name:
          RRP9, small subunit (SSU) processome component, homolog (yeast)
          Gene Aliases:
          55kDa, D19435, D9Wsu10e, Rnu3ip2, U3-55k
          Chromosome Location:
          Chr.9: 106477309 - 106485415 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00770070_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rrp9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145620.4 NP_663595.1 14 - 15 1379 137
          GenBank mRNA AF368232.1 - 14 - 15 1362 137
          AK076113.1 - 14 - 15 1381 137
          AK156199.1 - 13 - 14 1453 137
          BC014703.1 - 14 - 15 1379 4237895 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001350.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rrp9

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          biological_process
          RNA binding
          snoRNA binding
          U3 snoRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26803653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26803654 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 AF368232.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 AK076113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 AK156199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 BC014703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 NM_145620.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 AF368232.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 AK076113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 AK156199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 BC014703.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P518163 NM_145620.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38774 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 AF368232.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 AK076113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 AK156199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 BC014703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 NM_145620.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 AF368232.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 AK076113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 AK156199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 BC014703.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P518163 NM_145620.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 AF368232.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 AK076113.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 AK156199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 BC014703.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 NM_145620.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 AF368232.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 AK076113.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 AK156199.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 BC014703.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P518163 NM_145620.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26803657 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02022979 - -
          Whole Mouse Genome G4122A A_51_P518163 AF368232.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 AK076113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 AK156199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 BC014703.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 NM_145620.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 AF368232.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 AK076113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 AK156199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 BC014703.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518163 NM_145620.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117486 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116773 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02022979 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07853625 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07853626 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26803653 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26803654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26803649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07496853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07606712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08092595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07562672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08092598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07496855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07853614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08092609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07917258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26803679 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588534 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588527 - -
          Affymetrix HT MG-430 PM Array Plate 1451293_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10588522 - -
          Mouse Genome 430 2.0 Array 1451293_at - -
          Mouse Genome 430 2.0 Array 1451293_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27966_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27966_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900010565.mm.1 TC0900001350.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900010565.mm.1 NM_145620.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010565.mm.1 NM_145620.4 Refseq
          Murine Genome U74v2 Set 99954_at - -
          Murine Genome U74v2 Set 99954_PM_at - -
          Murine Genome U74v2 Set 169448_PM_r_at - -
          Murine Genome U74v2 Set 169448_r_at - -
          Affymetrix HT Mouse Genome 430A Array 1451293_at - -
          Affymetrix HT Mouse Genome 430A Array 1451293_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 674858 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 630451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223551 - -
          mouse-6 v1.1 (Illumina) scl36508.16.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 630451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36508.16.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223551 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223551 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36508.16.1_6-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0