Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00777269_mH
          See other PIN1 GE Assays ›
          Gene Symbol:
          Pin1
          Gene Name:
          protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1
          Gene Aliases:
          0610025L01Rik, D9Bwg1161e
          Chromosome Location:
          Chr.9: 20652130 - 20666584 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00777269_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pin1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023371.3 NP_075860.1 3 - 4 402 162
          GenBank mRNA AB009691.1 - 3 - 4 421 162
          AK002665.1 - 3 - 4 428 162
          AK003369.1 - 3 - 4 403 162
          AK003931.1 - 3 - 4 416 162
          AK054045.1 - 3 - 4 403 162
          AK139395.1 - 3 - 4 401 162
          AK150652.1 - 3 - 4 403 162
          AK160228.1 - 3 - 4 415 162
          BC038254.1 - 3 - 4 409 4971446 162

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000177.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pin1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          positive regulation of protein phosphorylation
          cell cycle
          negative regulation of transforming growth factor beta receptor signaling pathway
          regulation of cytokinesis
          positive regulation of protein dephosphorylation
          regulation of cell population proliferation
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          positive regulation of GTPase activity
          positive regulation of ubiquitin-protein transferase activity
          regulation of pathway-restricted SMAD protein phosphorylation
          positive regulation of cell growth involved in cardiac muscle cell development
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of cell motility
          peptidyl-prolyl cis-trans isomerase activity
          protein binding
          isomerase activity
          mitogen-activated protein kinase kinase binding
          GTPase activating protein binding
          phosphoserine residue binding
          phosphothreonine residue binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900001116.mm.1 TC0900000177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001116.mm.1 NM_023371.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001116.mm.1 NM_023371.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001117.mm.1 TC0900000177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001117.mm.1 NM_023371.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001117.mm.1 NM_023371.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001114.mm.1 TC0900000177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001114.mm.1 NM_023371.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001114.mm.1 NM_023371.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001113.mm.1 TC0900000177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001113.mm.1 NM_023371.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001113.mm.1 NM_023371.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001115.mm.1 TC0900000177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001115.mm.1 NM_023371.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001115.mm.1 NM_023371.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0