Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00780078_s1
          See other ENY2 GE Assays ›
          Gene Symbol:
          Eny2
          Gene Name:
          enhancer of yellow 2 homolog (Drosophila)
          Gene Aliases:
          1810057B09Rik, 6720481I12, DC6, Ey2
          Chromosome Location:
          Chr.15: 44428081 - 44438256 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00780078_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eny2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175009.3 NP_778174.1 5 1988 91
          GenBank mRNA AK032957.1 - 5 1988 91
          AK136334.1 - 1 1075 91
          AK162511.1 - 5 1983 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eny2

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA export from nucleus
          transport
          protein transport
          histone deubiquitination
          positive regulation of transcription, DNA-templated
          mRNA transport
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          transcription coactivator activity
          nuclear receptor coactivator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30180497 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30180498 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987302 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111419 AK162511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111419 BC048361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111419 AK032957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111419 AK160628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111419 AK007877.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 AK162511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 AK032957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 AK136334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 NM_175009.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 AK162511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 AK032957.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 AK136334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P653902 NM_175009.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707194 AK045765.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141422 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 AK162511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 AK032957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 AK136334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 NM_175009.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 AK162511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 AK032957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 AK136334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P653902 NM_175009.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111419 AK162511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111419 BC048361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111419 AK032957.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111419 AK160628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111419 AK007877.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30180511 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03023623 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08353081 - -
          Whole Mouse Genome G4122A A_52_P653902 AK162511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653902 AK032957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653902 AK136334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653902 NM_175009.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653902 AK162511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653902 AK032957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653902 AK136334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P653902 NM_175009.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P143576 BC048361.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135983 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127551 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03023623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11992665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12299047 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30180498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30180497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30180478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12268168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12442937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12442938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11992658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11992670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30180495 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10423965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423964 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10423970 - -
          Affymetrix HT MG-430 PM Array Plate 1460392_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1429411_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1429410_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429412_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456961_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457758_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10423963 - -
          Mouse Genome 430 2.0 Array 1457758_at - -
          Mouse Genome 430 2.0 Array 1429412_PM_at - -
          Mouse Genome 430 2.0 Array 1429411_PM_a_at - -
          Mouse Genome 430 2.0 Array 1429410_PM_at - -
          Mouse Genome 430 2.0 Array 1429411_a_at - -
          Mouse Genome 430 2.0 Array 1460392_a_at - -
          Mouse Genome 430 2.0 Array 1429410_at - -
          Mouse Genome 430 2.0 Array 1457758_PM_at - -
          Mouse Genome 430 2.0 Array 1456961_at - -
          Mouse Genome 430 2.0 Array 1460392_PM_a_at - -
          Mouse Genome 430 2.0 Array 1429412_at - -
          Mouse Genome 430 2.0 Array 1456961_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223527_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223527_PM_at - -
          Mouse Expression Set 430 1429410_at - -
          Mouse Expression Set 430 1429411_a_at - -
          Mouse Expression Set 430 1460392_PM_a_at - -
          Mouse Expression Set 430 1429411_PM_a_at - -
          Mouse Expression Set 430 1460392_a_at - -
          Mouse Expression Set 430 1429412_at - -
          Mouse Expression Set 430 1429410_PM_at - -
          Mouse Expression Set 430 1429412_PM_at - -
          Mouse Expression Set 430 1457758_PM_at - -
          Mouse Expression Set 430 1456961_at - -
          Mouse Expression Set 430 1456961_PM_at - -
          Mouse Expression Set 430 1457758_at - -
          Murine Genome U74v2 Set 98958_PM_at - -
          Murine Genome U74v2 Set 98958_at - -
          Murine Genome U74v2 Set 141046_PM_at - -
          Murine Genome U74v2 Set 141046_at - -
          Mouse Genome 430A 2.0 Array 1429412_at - -
          Mouse Genome 430A 2.0 Array 1429411_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1460392_a_at - -
          Mouse Genome 430A 2.0 Array 1460392_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1429410_PM_at - -
          Mouse Genome 430A 2.0 Array 1429412_PM_at - -
          Mouse Genome 430A 2.0 Array 1429410_at - -
          Mouse Genome 430A 2.0 Array 1429411_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429410_at - -
          Affymetrix HT Mouse Genome 430A Array 1429411_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429412_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429412_at - -
          Affymetrix HT Mouse Genome 430A Array 1460392_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1460392_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429410_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429411_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1457758_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456961_at - -
          Affymetrix HT Mouse Genome 430B Array 1457758_at - -
          Affymetrix HT Mouse Genome 430B Array 1456961_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 434366 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104230017 - -
          Illumina mouse-6 v1.0 expression beadchip 6110026 - -
          Illumina mouse-6 v1.0 expression beadchip 104230017 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2703033 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2703033 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254640 - -
          mouse-6 v1.1 (Illumina) scl47927.5_282-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110026 - -
          Illumina Sentrix mouse-6.v1 bead array 104230017 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47927.5_282-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104230017 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2703032 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2703032 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254640 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0