Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00782916_s1
          See other MRPL49 GE Assays ›
          Gene Symbol:
          Mrpl49
          Gene Name:
          mitochondrial ribosomal protein L49
          Gene Aliases:
          4833419D18Rik, Nof1
          Chromosome Location:
          Chr.19: 6053630 - 6057751 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00782916_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl49
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026246.3 NP_080522.1 4 731 148
          GenBank mRNA AK011273.1 - 3 588 148
          AK014723.1 - 4 732 148
          AK049794.1 - 4 732 148
          AK077397.1 - 4 731 148
          AK150572.1 - 4 736 148
          AK150715.1 - 4 736 148
          BC028956.1 - 4 721 5368499 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000999.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl49

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          molecular_function
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932435 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 AK150572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 AK150715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 AK014723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 NM_026246.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 AK150572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 AK150715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 AK014723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P261351 NM_026246.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957909 BC028956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957909 AK150572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957909 AK150715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957909 AK077397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957909 AK011273.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94465 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156878 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 AK150572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 AK150715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 AK014723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 NM_026246.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 AK150572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 AK150715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 AK014723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P261351 NM_026246.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957909 BC028956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957909 AK150572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957909 AK150715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957909 AK077397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957909 AK011273.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 AK150572.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 AK150715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 AK014723.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 NM_026246.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 AK150572.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 AK150715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 AK014723.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261351 NM_026246.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31932434 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156878 - -
          Whole Mouse Genome G4122A A_52_P485433 BC028956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485433 AK150572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485433 AK150715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485433 AK077397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485433 AK007712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 AK150572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 AK150715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 AK014723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 NM_026246.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 AK150572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 AK150715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 AK014723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261351 NM_026246.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484691 BC028956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484691 AK150572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484691 AK150715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484691 AK077397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484691 AK011273.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128119 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14192871 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14192860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14192864 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14192882 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14192880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31932436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31932360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14459425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14192865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14192882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14294060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14459427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14440658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14311289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31932417 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465338 - -
          Affymetrix HT MG-430 PM Array Plate 1423219_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1423218_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465336 - -
          Mouse Genome 430 2.0 Array 1423219_PM_a_at - -
          Mouse Genome 430 2.0 Array 1423218_a_at - -
          Mouse Genome 430 2.0 Array 1423219_a_at - -
          Mouse Genome 430 2.0 Array 1423218_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18120_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18120_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900009138.mm.1 TC1900000999.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009138.mm.1 NM_026246.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009138.mm.1 NM_026246.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009140.mm.1 TC1900000999.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009140.mm.1 NM_026246.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009140.mm.1 NM_026246.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009139.mm.1 TC1900000999.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009139.mm.1 NM_026246.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009139.mm.1 NM_026246.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009136.mm.1 TC1900000999.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009136.mm.1 NM_026246.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009136.mm.1 NM_026246.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009137.mm.1 TC1900000999.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009137.mm.1 NM_026246.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009137.mm.1 NM_026246.3 Refseq
          Mouse Expression Set 430 1423218_a_at - -
          Mouse Expression Set 430 1423219_PM_a_at - -
          Mouse Expression Set 430 1423219_a_at - -
          Mouse Expression Set 430 1423218_PM_a_at - -
          Murine Genome U74v2 Set 161597_PM_r_at - -
          Murine Genome U74v2 Set 161597_r_at - -
          Murine Genome U74v2 Set 94283_PM_at - -
          Murine Genome U74v2 Set 94283_at - -
          Mouse Genome 430A 2.0 Array 1423218_a_at - -
          Mouse Genome 430A 2.0 Array 1423218_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423219_a_at - -
          Mouse Genome 430A 2.0 Array 1423219_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423218_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423219_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423219_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423218_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 752552 - -
          ABI Mouse Genome Survey Microarray 897491 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5340156 - -
          Illumina mouse-6 v1.0 expression beadchip 460541 - -
          Illumina mouse-6 v1.0 expression beadchip 610162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460541 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2634295 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2628672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2701363 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2628672 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2701363 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2634295 - -
          mouse-6 v1.1 (Illumina) scl000455.1_29-S - -
          mouse-6 v1.1 (Illumina) scl018120.4_62-S - -
          mouse-6 v1.1 (Illumina) scl53524.2.2_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 460541 - -
          Illumina Sentrix mouse-6.v1 bead array 5340156 - -
          Illumina Sentrix mouse-6.v1 bead array 610162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000455.1_29-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53524.2.2_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018120.4_62-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610162 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460541 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2628672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2929031 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2701363 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2634295 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2634295 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2701363 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2929031 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2628672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018120.4_62-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0