Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00787782_s1
          See other SCP2D1 GE Assays ›
          Gene Symbol:
          Scp2d1
          Gene Name:
          SCP2 sterol-binding domain containing 1
          Gene Aliases:
          1700010M22Rik
          Chromosome Location:
          Chr.2: 144823666 - 144824415 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00787782_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scp2d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025490.2 NP_079766.3 1 698 126
          GenBank mRNA AK005847.1 - 1 698 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002155.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scp2d1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256549 AK015138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256549 AK015138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224412 AK005847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P224412 AK005847.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540656 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130229 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256549 AK015138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256549 AK015138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224412 AK005847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P224412 AK005847.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21709221 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130229 - -
          Whole Mouse Genome G4122A A_51_P256549 AK015138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256549 AK015138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224412 AK005847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P224412 AK005847.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130405 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02022421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02022422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02022423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21709255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01815187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02041591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02201993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01815185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01914295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01889311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01409967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02202000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01409973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01768125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21709221 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10476738 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476739 - -
          Affymetrix HT MG-430 PM Array Plate 1429465_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476737 - -
          Mouse Genome 430 2.0 Array 1429465_PM_at - -
          Mouse Genome 430 2.0 Array 1429465_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66328_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66328_at - -
          Affymetrix Mouse Clariom S Array PSR0200017037.mm.1 TC0200002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017037.mm.1 NM_025490.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017037.mm.1 NM_025490.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017038.mm.1 TC0200002155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017038.mm.1 NM_025490.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017038.mm.1 NM_025490.2 Refseq
          Mouse Expression Set 430 1429465_PM_at - -
          Mouse Expression Set 430 1429465_at - -
          Affymetrix HT Mouse Genome 430B Array 1429465_at - -
          Affymetrix HT Mouse Genome 430B Array 1429465_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 886722 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610441 - -
          MouseWG-6 v2 Expression BeadChip 6980722 - -
          Illumina mouse-6 v1.0 expression beadchip 3610441 - -
          Illumina mouse-6 v1.0 expression beadchip 6980722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2607828 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245353 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2607828 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245353 - -
          mouse-6 v1.1 (Illumina) scl0066328.1_325-S - -
          mouse-6 v1.1 (Illumina) scl20189.1.1_328-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610441 - -
          Illumina Sentrix mouse-6.v1 bead array 6980722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20189.1.1_328-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066328.1_325-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980722 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245353 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2607827 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2607827 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245353 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc