Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00803163_g1
          See other PIK3R1 GE Assays ›
          Gene Symbol:
          Pik3r1
          Gene Name:
          phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha)
          Gene Aliases:
          PI3K, p50alpha, p55alpha, p85alpha
          Chromosome Location:
          Chr.13: 101680761 - 101768217 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00803163_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pik3r1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024955.2 NP_001020126.1 8 - 9 1347 83
          NM_001077495.2 NP_001070963.1 14 - 15 2436 83
          GenBank mRNA AK046259.1 - 8 - 9 1350 83
          BC026146.1 - 14 - 15 2417 3979333 83
          BC051106.1 - 8 - 9 1047 5031261 83
          M60651.1 - 14 - 15 2371 83
          U50413.1 - 14 - 15 2389 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002553.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pik3r1

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular glucose homeostasis
          positive regulation of protein phosphorylation
          negative regulation of cell-matrix adhesion
          glucose metabolic process
          protein phosphorylation
          transport
          negative regulation of cell adhesion
          signal transduction
          insulin receptor signaling pathway
          extrinsic apoptotic signaling pathway via death domain receptors
          intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of heart rate
          positive regulation of gene expression
          negative regulation of muscle cell apoptotic process
          phosphatidylinositol 3-kinase signaling
          protein transport
          negative regulation of cell-cell adhesion
          B cell differentiation
          positive regulation of cell migration
          positive regulation of tumor necrosis factor production
          response to insulin
          cellular response to insulin stimulus
          positive regulation of RNA splicing
          cellular response to UV
          response to endoplasmic reticulum stress
          phosphatidylinositol-3-phosphate biosynthetic process
          negative regulation of apoptotic process
          regulation of phosphatidylinositol 3-kinase activity
          positive regulation of myoblast differentiation
          negative regulation of osteoclast differentiation
          negative regulation of blood pressure
          negative regulation of proteolysis
          positive regulation of transcription by RNA polymerase II
          phosphatidylinositol phosphate biosynthetic process
          insulin-like growth factor receptor signaling pathway
          negative regulation of smooth muscle cell proliferation
          protein stabilization
          response to glucocorticoid
          regulation of stress fiber assembly
          response to cAMP
          growth hormone receptor signaling pathway
          regulation of calcineurin-NFAT signaling cascade
          positive regulation of endoplasmic reticulum unfolded protein response
          regulation of protein localization to plasma membrane
          signaling receptor binding
          insulin receptor binding
          insulin-like growth factor receptor binding
          platelet-derived growth factor receptor binding
          neurotrophin TRKA receptor binding
          protein binding
          calmodulin binding
          protein C-terminus binding
          transcription factor binding
          1-phosphatidylinositol-3-kinase activity
          protein kinase binding
          protein phosphatase binding
          protein domain specific binding
          estrogen receptor binding
          receptor tyrosine kinase binding
          ubiquitin protein ligase binding
          phosphatidylinositol 3-kinase regulator activity
          phosphatidylinositol 3-kinase regulatory subunit binding
          ErbB-3 class receptor binding
          phosphatidylinositol 3-kinase binding
          insulin binding
          insulin receptor substrate binding
          1-phosphatidylinositol-3-kinase regulator activity
          protein heterodimerization activity
          ATPase binding
          phosphoprotein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300017401.mm.1 TC1300002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300017401.mm.1 NM_001024955.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300017401.mm.1 NM_001077495.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0