Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00804104_gH
          See other KIN GE Assays ›
          Gene Symbol:
          Kin
          Gene Name:
          antigenic determinant of rec-A protein
          Gene Aliases:
          Kin17
          Chromosome Location:
          Chr.2: 10080598 - 10092701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00804104_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kin
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025280.2 NP_079556.1 4 - 5 394 71
          GenBank mRNA AK009429.1 - 4 - 5 426 71
          AK167740.1 - 4 - 5 398 71
          BC028860.1 - 4 - 5 394 3673830 71
          X58472.1 - 4 - 5 399 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BC099685.1 - 4640 Chr. - - [-] on Build 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000082.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kin

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA repair
          DNA recombination
          mRNA processing
          cellular response to DNA damage stimulus
          DNA binding
          double-stranded DNA binding
          RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20998754 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20998755 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 BC058169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 X58472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 AK132548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 AK167740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 BC028860.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 BC058169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 X58472.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 AK132548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 BC028860.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436483 AK167740.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168264 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162291 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167986 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 BC058169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 X58472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 AK132548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 AK167740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 BC028860.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 BC058169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 X58472.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 AK132548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 AK167740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436483 BC028860.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20998777 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167986 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162291 - -
          Whole Mouse Genome G4122A A_51_P436483 BC058169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 X58472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 AK132548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 AK167740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 BC028860.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 BC058169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 X58472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 AK132548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 AK167740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436483 BC028860.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P455275 BC058169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P455275 X58472.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P455275 AK167740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P455275 BC028860.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P455275 NM_025280.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105103 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167986 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01537147 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01537145 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01537136 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20998755 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20998754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20998769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01690296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01185130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01506954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01236538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01653010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01537144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20998782 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469146 - -
          Affymetrix HT MG-430 PM Array Plate 1427317_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469145 - -
          Mouse Genome 430 2.0 Array 1427317_PM_at - -
          Mouse Genome 430 2.0 Array 1427317_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16588_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16588_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200000532.mm.1 TC0200000082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000532.mm.1 NM_025280.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000532.mm.1 NM_025280.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000535.mm.1 TC0200000082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000535.mm.1 NM_025280.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000535.mm.1 NM_025280.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000533.mm.1 TC0200000082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000533.mm.1 NM_025280.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000533.mm.1 NM_025280.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000534.mm.1 TC0200000082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000534.mm.1 NM_025280.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000534.mm.1 NM_025280.2 Refseq
          Mouse Expression Set 430 1427317_PM_at - -
          Mouse Expression Set 430 1427317_at - -
          Murine Genome U74v2 Set 160842_PM_at - -
          Murine Genome U74v2 Set 160842_at - -
          Mouse Genome 430A 2.0 Array 1427317_PM_at - -
          Mouse Genome 430A 2.0 Array 1427317_at - -
          Affymetrix HT Mouse Genome 430A Array 1427317_at - -
          Affymetrix HT Mouse Genome 430A Array 1427317_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6550014 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550014 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232589 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232589 - -
          mouse-6 v1.1 (Illumina) scl21299.5_175-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550014 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21299.5_175-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550014 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232589 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232589 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21299.5_175-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0