Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805006_m1
          See other KDM4A GE Assays ›
          Gene Symbol:
          Kdm4a
          Gene Name:
          lysine (K)-specific demethylase 4A
          Gene Aliases:
          D4Ertd222e, JHDM3A, Jmjd2, Jmjd2a, mKIAA0677
          Chromosome Location:
          Chr.4: 118136957 - 118180043 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805006_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kdm4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161823.1 NP_001155295.1 6 - 7 873 70
          NM_172382.2 NP_759014.2 6 - 7 873 70
          GenBank mRNA AK054095.1 - 6 - 7 837 70
          AK129187.1 - 6 - 7 728 70
          AK145066.1 - 6 - 7 859 70
          AK145947.1 - 6 - 7 795 70
          BC028866.1 - 6 - 7 776 4207552 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003385.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kdm4a

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of gene expression
          cardiac muscle hypertrophy in response to stress
          histone demethylation
          positive regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          negative regulation of astrocyte differentiation
          negative regulation of cell death
          histone H3-K36 demethylation
          sulfonate dioxygenase activity
          double-stranded DNA binding
          zinc ion binding
          oxidoreductase activity
          2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
          procollagen-proline dioxygenase activity
          ubiquitin protein ligase binding
          histone demethylase activity
          hypophosphite dioxygenase activity
          methylated histone binding
          DNA-N1-methyladenine dioxygenase activity
          metal ion binding
          dioxygenase activity
          histone H3-methyl-lysine-36 demethylase activity
          C-19 gibberellin 2-beta-dioxygenase activity
          C-20 gibberellin 2-beta-dioxygenase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400027311.mm.1 TC0400003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027311.mm.1 NM_001161823.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027311.mm.1 NM_001161823.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027311.mm.1 NM_172382.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027311.mm.1 NM_172382.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027312.mm.1 TC0400003385.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027312.mm.1 NM_001161823.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027312.mm.1 NM_001161823.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027312.mm.1 NM_172382.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027312.mm.1 NM_172382.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0